Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MKCKPNQTR TYDPEGFKKR AACLCFRSER EDEVLLVSSS RYPDRWIVPG GGMEPEEEPG GAAVREVYEE AGVKGKLGRL LGVFEQNQDR KHRTYVYVLT VTELLEDWED SVSIGRKREW FKVEDAIKVL QCHKPVHAEY LEKLKLGGSP TNGNSMAPSS PDSDP (1-164 aa encoded by BC009942)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3, beta; Diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP-3-alpha; DIPP-3-beta; DIPP3 beta; DIPP3-beta; hAps1; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 11; Nudix motif 10; Nudix motif 11; nudix-type motif 11; unnamed protein product
Gene Aliases: APS1; ASP1; DIPP3; Dipp3a; DIPP3B; DIPP3beta; hDIPP3beta; MNCb-1696; Nudt10; NUDT11
UniProt ID: (Human) Q96G61, (Mouse) P0C027
Entrez Gene ID: (Human) 55190, (Rat) 680248, (Mouse) 58242
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support