Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • UCP2 Antibodies

          Proteintech

          UCP2 Polyclonal Antibody

          60 References
          View all (31) UCP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UCP2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (22)
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UCP2 Antibody (11081-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human cervical cancer tissue slide using (Product # 11081-1-AP) (UCP2 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Western Blot (WB)
          UCP2 Antibody in Immunoprecipitation (IP)
          UCP2 Polyclonal Antibody

          Product Details

          11081-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 44 publications 44 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Immunoprecipitation (IP)

          1:500
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UCP2 Fusion Protein Ag1551 (1-309 aa encoded by BC011737)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody detect a band around 28-33 kDa corresponding to UCP2 protein in mouse liver, skeletal muscle and pancreas. Some upper and lower bands also were observed due to the dimer formation and degradation of the UCP2 protein. This antibody was raised against the full-length UCP2 protein and may cross-react with UCP3. It is notable that UCP2 is known to have a very short half-life less than 1 hour. It is strongly recommended that fresh-made lysate be used for western blot assays.

          Immunogen sequence: MVGFKATDV PPTATVKFLG AGTAACIADL ITFPLDTAKV RLQIQGESQG PVRATASAQY RGVMGTILTM VRTEGPRSLY NGLVAGLQRQ MSFASVRIGL YDSVKQFYTK GSEHASIGSR LLAGSTTGAL AVAVAQPTDV VKVRFQAQAR AGGGRRYQST VNAYKTIARE EGFRGLWKGT SPNVARNAIV NCAELVTYDL IKDALLKANL MTDDLPCHFT SAFGAGFCTT VIASPVDVVK TRYMNSALGQ YSSAGHCALT MLQKEGPRAF YKGFMPSFLR LGSWNVVMFV TYEQLKRALM AACTSREAPF (1-309 aa encoded by BC011737)

          Target Information

          UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Dicarboxylate carrier SLC25A8; Dicarboxylate carrier UCP2; Mitochondrial uncoupling protein 2; Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein; uncoupling protein 2 (mitochondrial, proton carrier); Uncoupling protein 2, mitochondrial

          View more View less

          Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH

          View more View less

          UniProt ID: (Human) P55851, (Rat) P56500, (Mouse) P70406

          View more View less

          Entrez Gene ID: (Human) 7351, (Rat) 54315, (Mouse) 22228

          View more View less

          Function(s)
          protein binding secondary active sulfate transmembrane transporter activity hydrogen ion transmembrane transporter activity chloride transmembrane transporter activity oxaloacetate transmembrane transporter activity malate transmembrane transporter activity L-aspartate transmembrane transporter activity antiporter activity oxidative phosphorylation uncoupler activity GDP binding protein homodimerization activity phosphate ion uniporter activity molecular_function transporter secondary carrier transporter
          Process(es)
          mitochondrial fission response to superoxide response to hypoxia glycolytic process glutamine metabolic process mitochondrial transport response to cold response to glucose inorganic anion transport C4-dicarboxylate transport long-chain fatty acid transport macrophage differentiation cellular response to insulin stimulus cellular response to hormone stimulus cellular response to amino acid starvation negative regulation of apoptotic process negative regulation of neuron apoptotic process regulation of mitochondrial membrane potential transmembrane transport negative regulation of insulin secretion involved in cellular response to glucose stimulus response to fatty acid L-aspartate transmembrane transport cellular response to lead ion cellular response to glucose stimulus malate transmembrane transport response to dexamethasone reactive oxygen species metabolic process liver regeneration negative regulation of calcium import into the mitochondrion positive regulation of cold-induced thermogenesis oxaloacetate(2-) transmembrane transport sulfate transmembrane transport chloride transmembrane transport hydrogen ion transmembrane transport mitochondrial transmembrane transport adaptive thermogenesis female pregnancy aging positive regulation of cell death transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline