Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • hnRNP A1 Antibodies

          Proteintech

          HNRNPA1 Polyclonal Antibody

          36 References
          View all (16) hnRNP A1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HNRNPA1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          HNRNPA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HNRNPA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HNRNPA1 Antibody (11176-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 4% PFA ) fixed HeLa cells using 11176-1-AP (HNRNPA1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HNRNPA1 Antibody in Immunocytochemistry (ICC/IF)
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA1 Antibody in Western Blot (WB)
          HNRNPA1 Antibody in Western Blot (WB)
          HNRNPA1 Antibody in Western Blot (WB)
          HNRNPA1 Polyclonal Antibody

          Product Details

          11176-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          View 26 publications 26 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 6 publications 6 publications

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig

          Published species

          Human, Mouse, Pig

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HNRNPA1 Fusion Protein Ag1656 (1-320 aa encoded by BC009600)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.16 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSKSESPKE PEQLRKLFIG GLSFETTDES LRSHFEQWGT LTDCVVMRDP NTKRSRGFGF VTYATVEEVD AAMNARPHKV DGRVVEPKRA VSREDSQRPG AHLTVKKIFV GGIKEDTEEH HLRDYFEQYG KIEVIEIMTD RGSGKKRGFA FVTFDDHDSV DKIVIQKYHT VNGHNCEVRK ALSKQEMASA SSSQRGRSGS GNFGGGRGGG FGGNDNFGRG GNFSGRGGFG GSRGGGGYGG SGDGYNGFGN DGSNFGGGGS YNDFGNYNNQ SSNFGPMKGG NFGGRSSGPY GGGGQYFAKP RNQGGYGGSS SSSSYGSGRR F (1-320 aa encoded by BC009600)

          Target Information

          HnRNP A1 belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre mRNAs in the nucleus and appear to influence pre mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. hnRNP A1 has two repeats of quasi RRM domains that bind to RNAs. It is one of the most abundant core proteins of hnRNP complexes and it is localized to the nucleoplasm. This protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re imported.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis secretory sperm binding protein; HDP-1; Helix-destabilizing protein; Heterogeneous nuclear ribonucleoprotein A1; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP A1-like 3; hnRNP core protein A1; hnRNP core protein A1-like 3; hnRNP I; HNRNP-I; HNRNPI; HNRPA1MGC102835; HNRPI; MGC102835; nuclear ribonucleoprotein particle A1 protein; pPTB; PTB; PTB-1; PTB-T; PTB2; PTB3; PTB4; putative heterogeneous nuclear ribonucleoprotein A1-like 3; RP11-78J21.1; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein; Single-strand-binding protein; Topoisomerase-inhibitor suppressed; unnamed protein product

          View more View less

          Gene Aliases: ALS19; ALS20; Fli-2; Hdp; HDP-1; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; MPD3; ROA1; Tis; UP 1

          View more View less

          UniProt ID: (Human) P09651, (Mouse) P49312

          View more View less

          Entrez Gene ID: (Pig) 768103, (Human) 3178, (Mouse) 15382

          View more View less

          Function(s)
          nucleic acid binding DNA binding single-stranded DNA binding RNA binding single-stranded RNA binding mRNA 3'-UTR binding protein binding protein domain specific binding miRNA binding pre-mRNA binding identical protein binding telomeric repeat-containing RNA binding G-rich strand telomeric DNA binding nucleotide binding
          Process(es)
          alternative mRNA splicing, via spliceosome mRNA processing transport RNA splicing mRNA transport regulation of alternative mRNA splicing, via spliceosome mRNA splicing, via spliceosome RNA export from nucleus negative regulation of telomere maintenance via telomerase positive regulation of telomere maintenance via telomerase cellular response to glucose starvation regulation of RNA splicing nuclear export nuclear import cellular response to sodium arsenite
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0