Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Folliculin Antibodies

          Proteintech

          FLCN Polyclonal Antibody

          6 References
          View all (9) Folliculin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FLCN Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          FLCN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FLCN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FLCN Antibody (11236-2-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human ovary tumor using 11236-2-AP (FLCN antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FLCN Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Western Blot (WB)
          FLCN Antibody in Immunoprecipitation (IP)
          FLCN Polyclonal Antibody

          Product Details

          11236-2-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FLCN Fusion Protein Ag1746 (1-342 aa encoded by BC015687)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.31 mg/mL

          Amount

          46.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody can recognize all the 3 isoforms of FLCN (38-45 kDa,64 kDa,20-26 kDa).

          Immunogen sequence: MNAIVALCH FCELHGPRTL FCTEVLHAPL PQGDGNEDSP GQGEQAEEEE GGIQMNSRMR AHSPAEGASV ESSSPGPKKS DMCEGCRSLA AGHPGYISHD KETSIKYVSH QHPSHPQLFS IVRQACVRSL SCEVCPGREG PIFFGDEQHG FVFSHTFFIK DSLARGFQRW YSIITIMMDR IYLINSWPFL LGKVRGIIDE LQGKALKVFE AEQFGCPQRA QRMNTAFTPF LHQRNGNAAR SLTSLTSDDN LWACLHTSFA WLLKACGSRL TEKLLEGAPT EDTLVQMEKL AGEAGVLLPG PWPGWPWGGT SCLLSWQESL REGNAALNQP RTSLREAHPP ISV (1-342 aa encoded by BC015687)

          Target Information

          This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; Folliculin; unnamed protein product

          View more View less

          Gene Aliases: AU014660; B430214A04Rik; BHD; DENND8B; FLCL; FLCN

          View more View less

          UniProt ID: (Human) Q8NFG4, (Mouse) Q8QZS3, (Rat) Q76JQ2

          View more View less

          Entrez Gene ID: (Human) 201163, (Mouse) 216805, (Rat) 303185

          View more View less

          Function(s)
          enzyme inhibitor activity GTPase activator activity protein binding enzyme binding macromolecular complex binding protein complex binding molecular_function
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development lysosome organization cell-cell junction assembly transforming growth factor beta receptor signaling pathway negative regulation of cell proliferation cellular response to starvation positive regulation of autophagy hemopoiesis positive regulation of transforming growth factor beta receptor signaling pathway SCF-dependent proteasomal ubiquitin-dependent protein catabolic process protein destabilization cellular response to nutrient levels TOR signaling regulation of TOR signaling negative regulation of TOR signaling positive regulation of TOR signaling lysosome localization cellular response to amino acid starvation negative regulation of Rho protein signal transduction intracellular signal transduction TORC1 signaling positive regulation of apoptotic process protein kinase B signaling negative regulation of glycolytic process negative regulation of transcription, DNA-templated regulation of Ras protein signal transduction epithelial cell proliferation negative regulation of epithelial cell proliferation negative regulation of protein kinase B signaling ERK1 and ERK2 cascade negative regulation of ERK1 and ERK2 cascade cell proliferation involved in kidney development energy homeostasis intrinsic apoptotic signaling pathway negative regulation of cold-induced thermogenesis negative regulation of protein localization to nucleus negative regulation of cell proliferation involved in kidney development negative regulation of post-translational protein modification negative regulation of intracellular signal transduction negative regulation of brown fat cell differentiation positive regulation of TORC1 signaling negative regulation of lysosome organization regulation of pro-B cell differentiation positive regulation of intrinsic apoptotic signaling pathway regulation of protein phosphorylation positive regulation of protein phosphorylation negative regulation of gene expression negative regulation of mitochondrion organization negative regulation of angiogenesis negative regulation of cell growth negative regulation of cell migration regulation of cytokinesis regulation of histone acetylation positive regulation of cell adhesion positive regulation of transcription from RNA polymerase II promoter negative regulation of cellular respiration negative regulation of mitochondrial DNA metabolic process negative regulation of muscle tissue development negative regulation of energy homeostasis negative regulation of ATP biosynthetic process biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline