Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EIQGLTTAH EQFKATLPDA DKERLAILGI HNEVSKIVQT YHVNMAGTNP YTTITPQEIN GKWDHVRQLV PRRDQALTEE HARQQHNERL RKQFGAQANV IGPWIQTKME EIGRISIEMH GTLEDQLSHL RQYEKSIVNY KPKIDQLEGD HQLIQEALIF DNKHTNYTME HIRVGWEQLL TTIARTINEV ENQILTRDAK GISQEQMNEF RASFNHFDRD HSGTLGPEEF KACLISLGYD IGNDPQGEAE FARIMSIVDP NRLGVVTFQA FIDFMSRETA DTDTADQVMA SFKILAGDKN YITMDELRRE LPPDQAEYCI ARMAPYTGPD SVPGALDYMS FSTALYGESD L (543-892 aa encoded by BC015766)
Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actinin 1 smooth muscle; alpha actinin 1a; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; epididymis secretory sperm binding protein; F-actin cross-linking protein; FLJ40884; FLJ54432; non-muscle alpha-actinin 1; Non-muscle alpha-actinin-1; unnamed protein product
Gene Aliases: 3110023F10Rik; ACTN1; Actn1a; BDPLT15
UniProt ID: (Human) P12814, (Rat) Q9Z1P2, (Mouse) Q7TPR4
Entrez Gene ID: (Human) 87, (Rat) 81634, (Mouse) 109711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support