Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • IGF2BP2 Antibodies

          Proteintech

          IGF2BP2 Polyclonal Antibody

          120 References
          View all (28) IGF2BP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IGF2BP2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (16)
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IGF2BP2 Antibody (11601-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human colon cancer tissue slide using (Product # 11601-1-AP) (IGF2BP2 antibody) at dilution of 1:1,000 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IGF2BP2 Antibody in Western Blot (WB)
          IGF2BP2 Antibody in Western Blot (WB)
          IGF2BP2 Antibody in Western Blot (WB)
          IGF2BP2 Antibody in Western Blot (WB)
          IGF2BP2 Antibody in Western Blot (WB)
          IGF2BP2 Antibody in Immunoprecipitation (IP)
          IGF2BP2 Polyclonal Antibody

          Product Details

          11601-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 56 publications 56 publications

          Immunohistochemistry (IHC)

          -
          View 18 publications 18 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 8 publications 8 publications

          RNA Immunoprecipitation (RIP)

          -
          View 29 publications 29 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IGF2BP2 Fusion Protein Ag2142 (1-341 aa encoded by BC021290)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody raise against part of N-terminus of human IGF2BP2.

          Immunogen sequence: MNKLYIGNL SPAVTADDLR QLFGDRKLPL AGQVLLKSGY AFVDYPDQNW AIRAIETLSG KVELHGKIME VDYSVSKKLR SRKIQIRNIP PHLQWEVLDG LLAQYGTVEN VEQVNTDTET AVVNVTYATR EEAKIAMEKL SGHQFENYSF KISYIPDEEV SSPSPPQRAQ RGDHSSREQG HAPGGTSQAR QIDFPLRILV PTQFVGAIIG KEGLTIKNIT KQTQSRVDIH RKENSGAAEK PVTIHATPEG TSEACRMILE IMQKEADETK LAEEIPLKIL AHNGLVGRLI GKEGRNLKKI EHETGTKITI SSLQDLSIYN PERTITVKGT VEACASAEIE IM (1-341 aa encoded by BC021290)

          Target Information

          IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Hepatocellular carcinoma autoantigen p62; IGF-II mRNA-binding protein 2; IGF2 mRNA-binding protein 2; IGF2BP2; RNA-binding protein; Insulin-like growth factor 2 mRNA-binding protein 2; insulin-like growth factor 2, binding protein 2; p62; unnamed protein product; VICKZ family member 2

          View more View less

          Gene Aliases: C330012H03Rik; IGF2BP2; IMP-2; IMP2; Neilsen; RGD1305614; VICKZ2

          View more View less

          UniProt ID: (Human) Q9Y6M1, (Mouse) Q5SF07

          View more View less

          Entrez Gene ID: (Human) 10644, (Mouse) 319765, (Rat) 303824

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA 3'-UTR binding protein binding translation regulator activity mRNA 5'-UTR binding N6-methyladenosine-containing RNA binding nucleotide binding mRNA binding poly(A) RNA binding RNA metabolism protein
          Process(es)
          regulation of cytokine production regulation of translation nervous system development anatomical structure morphogenesis negative regulation of translation negative regulation of proteasomal ubiquitin-dependent protein catabolic process proteasome-mediated ubiquitin-dependent protein catabolic process RNA stabilization mRNA transport CRD-mediated mRNA stabilization energy homeostasis cold-induced thermogenesis transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline