Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MHKEEHEVA VLGAPPSTIL PRSTVINIHS ETSVPDHVVW SLFNTLFLNW CCLGFIAFAY SVKSRDRKMV GDVTGAQAYA STAKCLNIWA LILGILMTIG FILLLVFGSV TVYHIMLQII QEKRGY (1-125 aa encoded by B C000897)
IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 27-Sep; CD225; Dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; ifitm-like protein 2; interferon induced transmembrane protein 2 like; Interferon-induced protein 17; Interferon-induced transmembrane protein 1; Interferon-inducible protein 9-27; Leu-13 antigen; Mil-2; Mouse ifitm-like protein 2; unnamed protein product
Gene Aliases: 1110036C17Rik; 9-27; CD225; DSPA2a; IFI17; IFITM1; LEU13; Mil-2; Mil2
UniProt ID: (Human) P13164, (Mouse) Q9D103
Entrez Gene ID: (Human) 8519, (Rat) 293618, (Mouse) 68713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support