Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ASEDVKEIF ARARNGKYRL LKISIENEQL VIGSYSQPSD SWDKDYDSFV LPLLEDKQPC YILFRLDSQN AQGYEWIFIA WSPDHSHVRQ KMLYAATRAT LKKEFGGGHI KDEVFGTVKE DVSLHGYKKY LLSQSSPAPL TAAEEELRQI KINEVQTDVG VDTKHQTLQG VAFPISREAF QALEKLNNRQ LNYVQLEIDI KNEIIILANT TNTELKDLPK RIPKDSARYH FFLYKHSHEG DYLESIVFIY SMPGYTCSIR ERMLYSSCKS RLLEIVERQL QMDVIRKIEI DNGDELTADF LYEEVHPKQH AHKQSFAKPK GPAGKRGIRR LIRGPAETEA TTD (1-343 aa encoded by BC022344)
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A6 protein tyrosine kinase; actin monomer-binding protein; Protein A6; Protein tyrosine kinase 9; PTK9; PTK9 protein tyrosine kinase 9; twinfilin, actin-binding protein, homolog 1; Twinfilin-1; unnamed protein product
Gene Aliases: A6; PTK9; TWF1; twinfilin
UniProt ID: (Human) Q12792, (Rat) Q5RJR2, (Mouse) Q91YR1
Entrez Gene ID: (Human) 5756, (Rat) 315265, (Mouse) 19230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support