Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TKPPPAKKA AESSSDSSDS DSSEDDEAPS KPAGTTKNSS NKPAVTTKSP AVKPAAAPKQ PVGGGQKLLT RKADSSSSEE ESSSSEEEKT KKMVATTKPK ATAKAALSLP AKQAPQGSRD SSSDSDSSSS EEEEEKTSKS AVKKKPQKVA GGAAPSKPAS AKKGKAESSN SSSSDDSSEE EEEKLKGKGS PRPQAPKANG TSALTAQNGK AAKNSEEEEE EKKKAAVVVS KSGSLKKRKQ NEAAKEAETP QAKKIKLQTP NTFPKRKKGE KRASSPFRRV REEEIEVDSR VADNSFDAKR GAAGDWGERA NQVLKFTKGK SFRHEKTKKK RGSYRGGSIS VQVNSIKFDS E (69-418 aa encoded by BC006769)
Related to nucleologenesis, may play a role in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It has intrinsic GTPase and ATPase activities. May play an important role in transcription catalyzed by RNA polymerase I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; Nopp140; Nucleolar 130 kDa protein; Nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphprotein 1; Nucleolar phosphoprotein p130; nucleolar protein p130; unnamed protein product
Gene Aliases: KIAA0035; NOLC1; NOPP130; NOPP140; NS5ATP13; P130; Srp40
UniProt ID: (Human) Q14978
Entrez Gene ID: (Human) 9221
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support