Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLEPQENGV IDLPDYEHVE DETFPPFPPP ASPERQDGEG TEPDEESGNG APVPVPPKRT VKRNIPKLDA QRLISERGLP ALRHVFDKAK FKGKGHEAED LKMLIRHMEH WAHRLFPKLQ FEDFIDRVEY LGSKKEVQTC LKRIRLDLPI LHEDFVSNND EVAENNEHDV TSTELDPFLT NLSESEMFAS ELSRSLTEEQ QQRIERNKQL ALERRQAKLL SNSQTLGNDM LMNTPRAHTV EEVNTDEDQK EESNGLNEDI LDNPCNDAIA NTLNEEETLL DQSFKNVQQQ LDATSRNITE AR (1-301 aa encoded by BC000870)
Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSM3 homolog; FLJ20516; TIMELESS-interacting protein; unnamed protein product
Gene Aliases: 1110005A05Rik; 1110018P21Rik; RGD1564084; TIPIN
UniProt ID: (Human) Q9BVW5, (Mouse) Q91WA1, (Rat) Q4QR88
Entrez Gene ID: (Human) 54962, (Mouse) 66131, (Rat) 363076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support