Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • LILRA4 Antibodies

          Invitrogen

          CD85g (ILT7) Monoclonal Antibody (eBio17G10.2 (17G10.2)), PE, eBioscience™

          1 Published Figure
          3 References
          View all (12) LILRA4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD85g (ILT7) Monoclonal Antibody (eBio17G10.2 (17G10.2)), PE, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          CD85g (ILT7) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD85g (ILT7) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD85g (ILT7) Antibody (12-5179-42) in Flow

          Staining of normal human peripheral blood cells with Anti-Human CD123 APC (Product # 17-1239-42) (left) or Anti-Human CD303a FITC (Product # 11-9818-42) (right) and Anti-Human CD85g (ILT7) PE (right). Qudrant lines demark isotype control (Product # 12-4714-81). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD85g (ILT7) Antibody in Flow Cytometry (Flow)
          CD85g (ILT7) Antibody in Flow Cytometry (Flow)

          Product Details

          12-5179-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.125 µg)/test
          View 2 publications 2 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG1

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio17G10.2 (17G10.2)

          Conjugate

          PE PE PE
          • APC 
          • Functional Grade
          • PerCP-Cyanine5.5
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10852713

          Product Specific Information

          Description: The eBio17G10.2 monoclonal antibody reacts with human immunoglobulin-like transcript 7 (ILT7), also known as CD85g. ILT7 is part of the leukocyte immunoglobulin-like receptor (LIR) gene family and associates with Fc epsilon R1 gamma to form a functional complex which transduces ITAM-mediated signals that negatively modulates TLR-induced type I IFN production by human plasmacytoid dendritic cells. ILT7 is expressed exclusively on plasmacytoid dendritic cells and is absent in monocyte derived DCs. Treatment with IL-3 or CpG causes a decrease in expression.

          Double staining of human PBMCs with eBio17G10.2 and anti-pDC marker BDCA2 yields virtually complete co-staining. Cross linking of ILT7 with eBio17G10.2 antibody causes reduced transcription of IFN alpha1, IFN alpha2, IFN alpha4 and IFN beta.

          Applications Reported: This eBio17G10.2 (17G10.2) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBio17G10.2 (17G10.2) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          ILT7 is a pDC-specific IgG-like transcript and is a member of the leukocyte immunoglobulin-like receptor gene family. This receptor, in complex with FcER1a, transduces ITAM-mediated signals which negatively modulate type I IFN production by pDC. ILT7 is thus a potential target to regulate Type 1 IFN production by pDC, which play a major role during viral infection, auto-immune diseases and anti-cancer immunity. Additionally, ILT7, along with CD123 and BDCA-2, can be used to identify or classify subsets of pDCalong with TLR expression and TLR mediated immune responses.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD85 antigen-like family member G; CD85g; ILT-7; immunoglobulin superfamily member; Immunoglobulin-like transcript 7; leucocyte Ig-like receptor A4; Leukocyte immunoglobulin-like receptor subfamily A member 4; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 4

          View more View less

          Gene Aliases: CD85g; ILT7; LILRA4

          View more View less

          UniProt ID: (Human) P59901

          View more View less

          Entrez Gene ID: (Human) 23547

          View more View less

          Function(s)
          receptor binding protein binding coreceptor activity inhibitory MHC class I receptor activity
          Process(es)
          immune system process immune response-regulating signaling pathway cell surface receptor signaling pathway cytokine-mediated signaling pathway negative regulation of interferon-alpha production negative regulation of tumor necrosis factor production negative regulation of toll-like receptor 7 signaling pathway negative regulation of toll-like receptor 9 signaling pathway Fc-epsilon receptor signaling pathway innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline