Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAEKRHTRD SEAQRLPDSF KDSPSKGLGP CGWILVAFSF LFTVITFPIS IWMCIKIIKE YERAIIFRLG RILQGGAKGP GLFFILPCTD SFIKVDMRTI SFDIPPQEIL TKDSVTISVD GVVYYRVQNA TLAVANITNA DSATRLLAQT TLRNVLGTKN LSQILSDREE IAHNMQSTLD DATDAWGIKV ERVEIKDVKL PVQLQRAMAA EAEASREARA KVIAAEGEMN ASRALKEASM VITEYPAALQ LRYLQTLTTI AAEKNSTIVF PLPIDMLQGI IGAKHSHLG (1-288 aa encoded by BC010703 )
Thought to regulate cation conductance. May regulate ACCN1 and ACCN3 gating.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Erythrocyte band 7 integral membrane protein; Erythrocyte membrane protein band 7.2; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte surface protein band 7.2; Protein 7.2b; STOM; Stomatin; unnamed protein product
Gene Aliases: BND7; EPB7; EPB72; STOM
UniProt ID: (Human) P27105
Entrez Gene ID: (Human) 2040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support