Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MENLMTSST LPPLFADEDG SKESNDLATT GLNHPEVPYS SGATSSTNNP EFVEDLSQGQ LLQSESSNAA EGNEQRHEDE QRSKRGGWSK GRKRKKPLRD SNAPKSPLTG YVRFMNERRE QLRAKRPEVP FPEITRMLGN EWSKLPPEEK QRYLDEADRD KERYMKELEQ YQKTEAYKVF SRKTQDRQKG KSHRQDAARQ ATHDHEKETE VKERSVFDIP IFTEEFLNHS KAREAELRQL RKSNMEFEER NAALQKHVES MRTAVEKLEV DVIQERSRNT VLQQHLETLR QVLTSSFASM PLPGSGETPT VDTIDSYMNR LHSIILANPQ DNENFIATVR EVVNRLDR (1-347 aa encoded by BC021959)
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10739; High mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; iBRAF; Inhibitor of BRAF35; unnamed protein product
Gene Aliases: 1200004E06Rik; 5730490E10Rik; HMG20A; HMGX1; HMGXB1; Ibraf; RGD1564760
UniProt ID: (Human) Q9NP66, (Mouse) Q9DC33
Entrez Gene ID: (Human) 10363, (Mouse) 66867, (Rat) 315689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support