Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PFLAYGEKG YGTVIPPQAS LVFHVLLIDV HNPKDAVQLE TLELPPGCVR RAGAGDFMRY HYNGSLMDGT LFDSSYSRNH TYNTYIGQGY IIPGMDQGLQ GACMGERRRI TIPPHLAYGE NGTGDKIPGS AVLIFNVHVI DFHNPADVVE IRTLSRPSET CNETTKLGDF VRYHYNCSLL DGTQLFTSHD YGAPQEATLG ANKVIEGLDT GLQGMCVGER RQLIVPPHLA HGESGARGVP GSAVLLFEVE LVSREDGLPT GYLFVWHKDP PANLFEDMDL NKDGEVPPEE FSTFIKAQVS EGKGRLMPGQ DPEKTIGDMF QNQDRNQDGK ITVDELKLKS DEDEERVHEE L (233-582 aa encoded by BC016467)
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa FK506-binding protein; 65 kDa FKBP; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP-10; FKBP-65; Immunophilin FKBP65; osteognesis imperfecta with congenital joint contractures; Peptidyl-prolyl cis-trans isomerase FKBP10; peptidyl-prolyl isomerase; Pfam PF00254; PPIase FKBP10; Rotamase; Telopeptide lysyl hydroxylase, bone-specific; unnamed protein product
Gene Aliases: AI325255; BRKS; BRKS1; FKBP-10; FKBP-65; Fkbp-rs; Fkbp-rs1; Fkbp1-rs; FKBP10; Fkbp6; FKBP65; Fkbprp; hFKBP65; OI11; OI6; PPIASE; PSEC0056; TLH1
UniProt ID: (Human) Q96AY3, (Mouse) Q61576
Entrez Gene ID: (Human) 60681, (Mouse) 14230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support