Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLSEVLLVS APGKVILHGE HAVVHGKVAL AVSLNLRTFL RLQPHSNGKV DLSLPNIGIK RAWDVARLQS LDTSFLEQGD VTTPTSEQVE KLKEVAGLPD DCAVTERLAV LAFLYLYLSI CRKQRALPSL DIVVWSELPP GAGLGSSAAY SVCLAAALLT VCEEIPNPLK DGDCVNRWTK EDLELINKWA FQGERMIHGN PSGVDNAVST WGGALRYHQG KISSLKRSPA LQILLTNTKV PRNTRALVAG VRNRLLKFPE IVAPLLTSID AISLECERVL GEMGEAPAPE QYLVLEELID MNQHHLNALG VGHASLDQLC QVTRARGLHS KLTGAGGGGC GITLLKPGLE QPEVEATKQA LTSCGFDCLE TSIGAPGVSI HSATSLDSRV QQALDGL (1-396 aa encoded by BC016140)
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Mevalonate kinase; mevalonate kinase 1; MK; unnamed protein product
Gene Aliases: 2310010A05Rik; AI256848; AI414037; LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426, (Mouse) Q9R008, (Rat) P17256
Entrez Gene ID: (Human) 4598, (Mouse) 17855, (Rat) 81727
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support