Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CXCL1 (GRO alpha) Antibodies

          Proteintech

          GRO alpha Polyclonal Antibody

          45 References
          View all (32) CXCL1 (GRO alpha) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GRO alpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GRO alpha Antibody (12335-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human gliomas tissue slide using (Product # 12335-1-AP) (GRO alpha antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GRO alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GRO alpha Polyclonal Antibody

          Product Details

          12335-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          -
          View 19 publications 19 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 12 publications 12 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GRO alpha Fusion Protein Ag2917 (1-107 aa encoded by BC011976)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MARAALSAA PSNPRLLRVA LLLLLLVAAG RRAAGASVAT ELRCQCLQTL QGIHPKNIQS VNVKSPGPHC AQTEVIATLK NGRKACLNPA SPIVKKIIEK MLNSDKSN (1-107 aa encoded by BC011976)

          Target Information

          CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 1; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); CXC; CXC motif chemokine; CXCL; CXCL-1; fibroblast secretory protein; GRO a; gro protein; GRO α; GRO-alpha(1-73); GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROalpha; Growth-regulated alpha protein; GROα; KC/GR)-alpha; KC/GRO-alpha; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; NAP-3; NAP3; Neutrophil-activating protein 3; Platelet-derived growth factor-inducible protein KC; Secretory protein N51; unnamed protein product

          View more View less

          Gene Aliases: CXCL1; FSP; GRO; GRO1; GROA; KC; MGSA; MGSA-a; N51; NAP-3; SCYB1

          View more View less

          UniProt ID: (Human) P09341, (Mouse) P12850

          View more View less

          Entrez Gene ID: (Human) 2919, (Mouse) 14825

          View more View less

          Function(s)
          receptor binding cytokine activity protein binding chemokine activity enzyme activator activity growth factor activity CXCR chemokine receptor binding chemokine
          Process(es)
          chemotaxis defense response inflammatory response immune response signal transduction G-protein coupled receptor signaling pathway nervous system development negative regulation of cell proliferation actin cytoskeleton organization killing of cells of other organism intracellular signal transduction cell chemotaxis antimicrobial humoral immune response mediated by antimicrobial peptide response to molecule of bacterial origin acute inflammatory response positive regulation of leukocyte chemotaxis positive regulation of cytosolic calcium ion concentration positive regulation of sodium ion transport neutrophil chemotaxis response to lipopolysaccharide positive regulation of superoxide anion generation positive regulation of potassium ion transport chemokine-mediated signaling pathway positive regulation of neutrophil mediated killing of fungus positive regulation of hematopoietic stem cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline