Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TSMLLGAQK PLSQAVKLML EGITASRNTL AKFLPGKRKQ LREVRESIRF LRQVGRDWVQ RRREALKRGE EVPADILTQI LKAEEGAQDD EGLLDNFVTF FIAGHETSAN HLAFTVMELS RQPEIVARLQ AEVDEVIGSK RYLDFEDLGR LQYLSQVLKE SLRLYPPAWG TFRLLEEETL IDGVRVPGNT PLLFSTYVMG RMDTYFEDPL TFNPDRFGPG APKPRFTYFP FSLGHRSCIG QQFAQMEVKV VMAKLLQRLE FRLVPGQRFG LQEQATLKPL DPVLCTLRPR GWQPAPPPPP C (201-500 aa encoded by BC022539)
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is expressed in the brain, where it converts cholesterol to 24S-hydroxycholesterol. While cholesterol cannot pass the blood-brain barrier, 24S-hydroxycholesterol can be secreted in the brain into the circulation to be returned to the liver for catabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CH24H; Cholesterol 24-hydroxylase; Cholesterol 24-monooxygenase; Cholesterol 24S-hydroxylase; cholestrol 24-hydroxylase; Cytochrome P450 46A1; cytochrome P450, 46 (cholesterol 24-hydroxylase); cytochrome P450, 46a1 (cholesterol 24-hydroxylase); cytochrome P450, family 46, subfamily A, polypeptide 1; cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase); unnamed protein product
Gene Aliases: CP46; CYP46; CYP46A1
UniProt ID: (Human) Q9Y6A2, (Mouse) Q9WVK8
Entrez Gene ID: (Human) 10858, (Rat) 362782, (Mouse) 13116
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support