Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Adenylate Kinase 3 Antibodies

          Proteintech

          AK3 Polyclonal Antibody

          2 References
          View all (26) Adenylate Kinase 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AK3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (27)
          AK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AK3 Antibody (12562-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using (Product # 12562-1-AP) (AK3 antibody) at dilution of 1:50 and CoraLite488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AK3 Antibody in Immunocytochemistry (ICC/IF)
          AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Western Blot (WB)
          AK3 Antibody in Immunoprecipitation (IP)
          AK3 Polyclonal Antibody

          Product Details

          12562-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AK3 Fusion Protein Ag3146 (1-227 aa encoded by BC013771)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.32 mg/mL

          Amount

          48 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGASARLLR AVIMGAPGSG KGTVSSRITT HFELKHLSSG DLLRDNMLRG TEIGVLAKAF IDQGKLIPDD VMTRLALHEL KNLTQYSWLL DGFPRTLPQA EALDRAYQID TVINLNVPFE VIKQRLTARW IHPASGRVYN IEFNPPKTVG IDDLTGEPLI QREDDKPETV IKRLKAYEDQ TKPVLEYYQK KGVLETFSGT ETNKIWPYVY AFLQTKVPQR SQKASVTP (1-227 aa encoded by BC013771)

          Target Information

          This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Adenylate kinase 3; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 3 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 6, adenylate kinase 3 like 1; Adenylate kinase isozyme 3; AK 3; AK 3 {ECO:0000255|HAMAP-Rule:MF_03169}; AK4; GTP-ATPPT; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03169}; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial; GTPAMPPT; MGAPT protein; unnamed protein product

          View more View less

          Gene Aliases: AA407498; AI506714; AK-3; AK3; Ak3l; AK3L1; AK6; AKL3L; AKL3L1; FIX

          View more View less

          UniProt ID: (Human) Q9UIJ7, (Rat) P29411, (Mouse) Q9WTP7

          View more View less

          Entrez Gene ID: (Human) 50808, (Rat) 26956, (Mouse) 56248

          View more View less

          Function(s)
          nucleotide binding adenylate kinase activity protein binding ATP binding GTP binding kinase activity transferase activity phosphotransferase activity, phosphate group as acceptor ligase activity nucleobase-containing compound kinase activity nucleoside triphosphate adenylate kinase activity identical protein binding nucleotide kinase
          Process(es)
          nucleobase-containing compound metabolic process ADP biosynthetic process blood coagulation nucleotide metabolic process nucleoside monophosphate metabolic process nucleoside triphosphate biosynthetic process AMP metabolic process GTP metabolic process ITP metabolic process UTP metabolic process liver development AMP phosphorylation muscle organ development cerebellum development olfactory bulb development response to starvation ATP metabolic process dATP metabolic process nucleotide phosphorylation protein homooligomerization phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline