Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BBS4 Antibodies

          Proteintech

          BBS4 Polyclonal Antibody

          19 References
          View all (16) BBS4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BBS4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          BBS4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BBS4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BBS4 Antibody (12766-1-AP) in ICC/IF

          IF result (cytoplasm and the length of cilia cilia stain) of anti-BBS4 (12766-1-AP; 1:50) with serum-starved hTERT-RPE1 cell (MeOH fixed) by Dr. Moshe Kim. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BBS4 Antibody in Immunocytochemistry (ICC/IF)
          BBS4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS4 Antibody in Western Blot (WB)
          BBS4 Antibody in Western Blot (WB)
          BBS4 Antibody in Western Blot (WB)
          BBS4 Antibody in Western Blot (WB)
          BBS4 Antibody in Western Blot (WB)
          BBS4 Polyclonal Antibody

          Product Details

          12766-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BBS4 Fusion Protein Ag3430 (1-351 aa encoded by BC027624)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.14 mg/mL

          Amount

          21 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MAEERVATR TQFPVSTESQ KPRQKKAPEF PILEKQNWLI HLHYIRKDYE ACKAVIKEQL QETQGLCEYA IYVQALIFRL EGNIQESLEL FQTCAVLSPQ SADNLKQVAR SLFLLGKHKA AIEVYNEAAK LNQKDWEISH NLGVCYIYLK QFNKAQDQLH NALNLNRHDL TYIMLGKIHL LEGDLDKAIE VYKKAVEFSP ENTELLTTLG LLYLQLGIYQ KAFEHLGNAL TYDPTNYKAI LAAGSMMQTH GDFDVALTKY RVVACAVPES PPLWNNIGMC FFGKKKYVAA ISCLKRANYL APFDWKILYN LGLVHLTMQQ YASAFHFLSA AINFQPKMGE LYMLLAVALT NL (1-351 aa encoded by BC027624)

          Target Information

          BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog; BBS4; BBSome complex member BBS4; unnamed protein product

          View more View less

          Gene Aliases: AW537059; AW742241; BBS4; D9Ertd464e

          View more View less

          UniProt ID: (Human) Q96RK4, (Mouse) Q8C1Z7

          View more View less

          Entrez Gene ID: (Human) 585, (Mouse) 102774, (Rat) 300754

          View more View less

          Function(s)
          protein binding protein binding, bridging dynactin binding alpha-tubulin binding beta-tubulin binding RNA polymerase II sequence-specific DNA binding transcription factor binding RNA polymerase II repressing transcription factor binding microtubule motor activity molecular_function
          Process(es)
          microtubule cytoskeleton organization mitotic cytokinesis neuron migration leukocyte homeostasis B cell homeostasis neural tube closure retina homeostasis heart looping lymphocyte homeostasis myeloid cell homeostasis negative regulation of systemic arterial blood pressure centrosome cycle spermatid development sensory perception visual perception sensory perception of smell protein localization gene expression negative regulation of gene expression protein transport Wnt signaling pathway dendrite development regulation of lipid metabolic process ventricular system development striatum development hippocampus development cerebral cortex development cell projection organization adult behavior negative regulation of actin filament polymerization melanosome transport regulation of cytokinesis leptin-mediated signaling pathway protein localization to organelle erythrocyte homeostasis microtubule anchoring at centrosome social behavior photoreceptor cell outer segment organization negative regulation of appetite by leptin-mediated signaling pathway positive regulation of multicellular organism growth response to leptin fat cell differentiation photoreceptor cell maintenance positive regulation of cilium assembly retinal rod cell development brain morphogenesis homeostasis of number of cells sensory processing maintenance of protein location in nucleus regulation of stress fiber assembly cilium morphogenesis regulation of cilium beat frequency involved in ciliary motility face development adipose tissue development fat pad development protein localization to cilium protein localization to centrosome sperm flagellum assembly regulation of nonmotile primary cilium assembly protein localization to photoreceptor outer segment non-motile cilium assembly transport metabolic process negative regulation of GTPase activity nonmotile primary cilium assembly cilium assembly centrosome organization biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline