Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human HDAC2.
Immunogen sequence: TDRVMTVSF HKYGEYFPGT GDLRDIGAGK GKYYAVNFPM RDGIDDESYG QIFKPIISKV MEMYQPSAVV LQCGADSLSG DRLGCFNLTV KGHAKCVEVV KTFNLPLLML GGGGYTIRNV ARCWTHETAV ALDCEIPNEL PYNDYFEYFG PDFKLHISPS NMTNQNTPEY MEKIKQRLFE NLRMLPHAPG VQMQAIPEDA VHEDSGDEDG EDPDKRISIR ASDKRIACDE EFSDSEDEGE GGRRNVADHK KGAKKARIEE DKKETEDKKT DVKEEDKSKD NSGEKTDTKG TKSEQLSNP (274-571 aa encoded by BC031055)
HDAC2 belongs to the histone deacetylase family. Histone deacetylases act via the formation of large multiprotein complexes and are responsible for the deacetylation of lysine residues at the N-terminal regions of core histones (H2A, H2B, H3 and H4). This protein forms transcriptional repressor complexes by associating with many different proteins, including YY1, a mammalian zinc-finger transcription factor. Thus, it plays an important role in transcriptional regulation, cell cycle progression and developmental events. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD2; hdac 2; hdac-2; Histone deacetylase 2; Protein deacylase HDAC2; transcriptional regulator homolog RPD3; unnamed protein product; YY1 transcription factor-binding protein; YY1-associated factor 1
Gene Aliases: D10Wsu179e; HD2; HDAC2; KDAC2; mRPD3; RPD3; YAF1; Yy1bp
UniProt ID: (Human) Q92769, (Mouse) P70288
Entrez Gene ID: (Human) 3066, (Mouse) 15182, (Rat) 84577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support