Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AIGGAVQDW ATFAVGPGHG VQLPSGRLLV PAYTYRVDRR ECFGKICRTS PHSFAFYSDD HGRTWRCGGL VPNLRSGECQ LAAVDGGQAG SFLYCNARSP LGSRVQALST DEGTSFLPAE RVASLPETAW GCQGSIVGFP APAPNRPRDD SWSVGPGSPL QPPLLGPGVH EPPEEAAVDP RGGQVPGGPF SRLQPRGDGP RQPGPRPGVS GDVGSWTLAL PMPFAAPPQS PTWLLYSHPV GRRARLHMGI RLSQSPLDPR SWTEPWVIYE GPSGYSDLAS IGPAPEGGLV FACLYESGAR TSYDEISFCT FSLREVLENV PASPKPPNLG DKPRGCCWPS (158-496 aa encoded by BC012899)
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-acetyl-alpha-neuraminidase 4; neuraminidase 4 (sialidase); Sialidase-4; unnamed protein product
Gene Aliases: LP5125; NEU4
UniProt ID: (Human) Q8WWR8
Entrez Gene ID: (Human) 129807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support