Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CD49f (Integrin alpha 6) Antibodies

          Invitrogen

          CD49f (Integrin alpha 6) Monoclonal Antibody (eBioGoH3 (GoH3)), eBioscience™

          59 Published Figures
          71 References
          View all (39) CD49f (Integrin alpha 6) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD49f (Integrin alpha 6) Monoclonal Antibody (eBioGoH3 (GoH3)), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (59)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 60

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD49f (Integrin alpha 6) Antibody (14-0495-82) in Flow

          Staining of normal human peripheral blood cells with 0.25 µg of Rat IgG2a Isotype Control Purified (Product # 14-4321-82) (open histogram) or 0.25 µg of Anti-Human/Mouse CD49f (Integrin alpha 6) Purified (filled histogram) followed by Anti-Rat IgG PE (Product # 12-4822). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunocytochemistry (ICC/IF)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Western Blot, Flow Cytometry (WB, Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Neutralization (Neu)

          Product Details

          14-0495-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 49 publications 49 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Neutralization (Neu)

          -
          View 2 publications 2 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioGoH3 (GoH3)

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_891480

          Product Specific Information

          Description: The eBioGoH3 monoclonal antibody reacts with mouse and human CD49f, also known as integrin alpha 6, very late activation antigen 6 (VLA-6 alpha chain), and platelet gpIc. CD49f is a 120 kD transmembrane protein. CD49f associates with CD29, the integrin beta 1 chain, to form the VLA-6 complex; CD49f also associates with CD104, the integrin beta 4 chain, to form the alpha 6 beta 4 complex. CD49f is expressed primarily on T cells, monocytes, platelets, epithelial and endothelial cells. CD49f expression has also been found on germinal center B cells. The eBioGoH3 antibody is cross-reactive to integrin alpha 6 on human, mouse and bovine cells. This antibody has also been reported to have functional activity in blocking the binding of integrin alpha 6 to laminin.

          Applications Reported: This eBioGoH3 (GoH3) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections.

          Applications Tested: This eBioGoH3 (GoH3) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CDf (Integrin alpha-6, ITGA6) is the integrin alpha chain alpha 6 protein. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signaling. Two transcript variants encoding different isoforms have been found for CDf. Functionally, integrins are cell surface receptors that interact with the extracellular matrix and mediate intracellular signals that affect cellular shape, mobility, and progression through the cell cycle in response to the extracellular matrix. Integrin receptors are composed of alpha and beta subunits, and form structural and functional linkages between the ECM (extra-cellular matrix) and intracellular cytoskeletal linker proteins. Signaling mediated by Intergrin/ECM interactions are also integrated with cellular responses to growth factor signaling to regulate cellular proliferation, cytoskeletal reorganization and other responses necessary for cellular survival. CDf is predominantly expressed by epithelia, and plays a critical structural role in the hemidesmosome. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD49 antigen-like family member F; CD49f; DKFZp686J01244; extracellular matrix receptor subunit; FLJ18737; Integrin alpha-6; integrin alpha6B; integrin, alpha 6; unnamed protein product; very late activation protein 6; VLA-6

          View more View less

          Gene Aliases: 5033401O05Rik; AI115430; CD49f; ITGA6; ITGA6A; ITGA6B; JEB6; VLA-6

          View more View less

          UniProt ID: (Human) P23229, (Mouse) Q61739

          View more View less

          Entrez Gene ID: (Human) 3655, (Mouse) 16403

          View more View less

          Function(s)
          protein binding insulin-like growth factor I binding signaling receptor activity neuregulin binding cadherin binding metal ion binding integrin binding protein complex binding laminin binding integrin
          Process(es)
          cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway ectodermal cell differentiation positive regulation of neuron projection development positive regulation of cell migration cell-substrate adhesion nail development positive regulation of apoptotic process positive regulation of GTPase activity skin morphogenesis positive regulation of transcription from RNA polymerase II promoter leukocyte migration cell-cell adhesion negative regulation of extrinsic apoptotic signaling pathway positive regulation of cell-substrate adhesion single organismal cell-cell adhesion positive regulation of cell-cell adhesion cellular response to extracellular stimulus cell adhesion mediated by integrin positive regulation of phosphorylation odontogenesis of dentin-containing tooth skin development filopodium assembly digestive tract development brown fat cell differentiation renal system development amelogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline