Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD49b (Integrin alpha 2) Antibodies

          Invitrogen

          CD49b (Integrin alpha 2) Monoclonal Antibody (eBioY418 (Y418)), eBioscience™

          Advanced Verification
          1 Reference
          View all (54) CD49b (Integrin alpha 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD49b (Integrin alpha 2) Monoclonal Antibody (eBioY418 (Y418)), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          CD49b (Integrin alpha 2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CD49b (Integrin alpha 2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD49b (Integrin alpha 2) Antibody (14-0498-80) in ICC/IF

          Immunofluorescence analysis of CD49b/ITGA2 was performed using 70% confluent log phase U-2 OS cells. The cells were fixed with 4% paraformaldehyde for 10 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with CD49b/ITGA2 Mouse Monoclonal Antibody (Product # 14-0498-80) at 5 µg/mL in 0.1% BSA, incubated at 4 degree Celsius overnight and then labeled with Goat anti-Mouse I... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD49b (Integrin alpha 2) Antibody in Immunocytochemistry (ICC/IF)
          CD49b (Integrin alpha 2) Antibody in Western Blot (WB)
          CD49b (Integrin alpha 2) Antibody

          Product Details

          14-0498-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.25 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          -

          Flow Cytometry (Flow)

          0.25 µg/test
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioY418 (Y418)

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FITC
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467289

          Product Specific Information

          Description: The eBioY418 (Y418) monoclonal antibody reacts with human CD49b, the 150-kDa, integrin alpha 2 subunit. The complex of CD49b non-covalently associated with integrin beta 1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by lymphocytes, activated T cells, monocytes and platelets.

          Applications Reported: This eBioY418 (Y418) antibody has been reported for use in flow cytometric analysis. (Fluorochrome-conjugated eBioY418 is recommended for use in flow cytometry.).

          Applications Tested: This eBioY418 (Y418) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha 2 integrin; alpha 2 subunit of VLA-2 receptor; BDPLT9; CD49 antigen-like family member B; CD49b; Collagen receptor; glycoprotein Ia; GPIa; human platelet alloantigen 5; human platelet alloantigen system 5; Integrin alpha-2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); platelet antigen Br; platelet glycoprotein GPIa; Platelet membrane glycoprotein Ia; unnamed protein product; very late activation protein 2 receptor, alpha-2 subunit; very late activation receptor alpha-2 subunit; very late activation receptor subunit alpha-2; VLA-2 subunit alpha; VLA2 receptor

          View more View less

          Gene Aliases: BR; CD49B; GPIa; HPA-5; ITGA2; VLA-2; VLAA2

          View more View less

          UniProt ID: (Human) P17301

          View more View less

          Entrez Gene ID: (Human) 3673

          View more View less

          Function(s)
          beta-amyloid binding virus receptor activity integrin binding protein binding collagen binding signaling receptor activity collagen receptor activity laminin binding heparan sulfate proteoglycan binding macromolecular complex binding metal ion binding collagen binding involved in cell-matrix adhesion
          Process(es)
          response to hypoxia positive regulation of leukocyte migration substrate-dependent cell migration hypotonic response cell adhesion cell-matrix adhesion integrin-mediated signaling pathway female pregnancy blood coagulation response to xenobiotic stimulus organ morphogenesis positive regulation of epithelial cell migration response to amine response to muscle activity positive regulation of smooth muscle cell migration extracellular matrix organization mammary gland development positive regulation of cell projection organization cell-substrate adhesion positive regulation of collagen biosynthetic process response to L-ascorbic acid cell adhesion mediated by integrin collagen-activated signaling pathway skin morphogenesis positive regulation of translation positive regulation of cell adhesion positive regulation of smooth muscle contraction viral entry into host cell focal adhesion assembly mesodermal cell differentiation positive regulation of smooth muscle cell proliferation positive regulation of positive chemotaxis detection of mechanical stimulus involved in sensory perception of pain positive regulation of transmission of nerve impulse positive regulation of phagocytosis, engulfment hepatocyte differentiation response to parathyroid hormone cellular response to mechanical stimulus cellular response to estradiol stimulus cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline