Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD105 (Endoglin) Antibodies

          Invitrogen

          CD105 (Endoglin) Monoclonal Antibody (SN6), eBioscience™

          66 Published Figures
          68 References
          View all (151) CD105 (Endoglin) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD105 (Endoglin) Monoclonal Antibody (SN6), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (66)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 67

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD105 (Endoglin) Antibody (14-1057-82) in Flow

          Staining of U937 cell line with 0.5 µg of Mouse IgG1 kappa Isotype Control Purified (Product # 14-4714-82) (open histogram) or Anti-Human CD105 (Endoglin) Purified (filled histogram) followed by F (ab')2 Anti-Mouse IgG PE (Product # 12-4012). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Immunocytochemistry (ICC/IF)
          CD105 (Endoglin) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD105 (Endoglin) Antibody in Immunohistochemistry (IHC)
          CD105 (Endoglin) Antibody in Western Blot (WB)
          CD105 (Endoglin) Antibody in Western Blot (WB)
          CD105 (Endoglin) Antibody in Western Blot (WB)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)
          CD105 (Endoglin) Antibody in Flow Cytometry (Flow)

          Product Details

          14-1057-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 60 publications 60 publications

          GST Pull Down (GST-PD)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Cat, Fruit fly, Horse, Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SN6

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • APC-eFluor 780
          • Brilliant UV 661
          • Brilliant Violet 605
          • eFluor 450
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467417

          Product Specific Information

          Description: The SN6 monoclonal antibody reacts with human CD105, also known as Endoglin. CD105, an approximately 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells and some bone marrow cells and activated macrophages. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.

          Applications Reported: The SN6 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The SN6 antibody has been tested by flow cytometric analysis of U937 cell line. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD105; CD105 antigen; circulating isoform of endoglin TGF-beta receptor type III; Endoglin; endoglin precursor; ORW; soluble endoglin; unnamed protein product

          View more View less

          Gene Aliases: END; ENG; HHT1; ORW1

          View more View less

          UniProt ID: (Human) P17813

          View more View less

          Entrez Gene ID: (Human) 2022

          View more View less

          Function(s)
          transmembrane signaling receptor activity type II transforming growth factor beta receptor binding protein binding galactose binding glycosaminoglycan binding coreceptor activity receptor activator activity type I transforming growth factor beta receptor binding BMP binding identical protein binding protein homodimerization activity activin binding transforming growth factor beta binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis patterning of blood vessels vasculogenesis response to hypoxia negative regulation of endothelial cell proliferation heart looping sprouting angiogenesis positive regulation of systemic arterial blood pressure outflow tract septum morphogenesis epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion morphogenesis cardiac ventricle morphogenesis cardiac atrium morphogenesis ventricular trabecula myocardium morphogenesis cell migration involved in endocardial cushion formation regulation of transcription, DNA-templated cell adhesion transforming growth factor beta receptor signaling pathway heart development negative regulation of gene expression regulation of cardiac muscle cell apoptotic process cell migration regulation of transforming growth factor beta receptor signaling pathway central nervous system vasculogenesis extracellular matrix disassembly regulation of cell adhesion negative regulation of cell migration BMP signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of BMP signaling pathway dorsal aorta morphogenesis wound healing regulation of cell proliferation positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter smooth muscle tissue development artery morphogenesis venous blood vessel morphogenesis cell motility positive regulation of protein kinase B signaling atrial cardiac muscle tissue morphogenesis cell chemotaxis positive regulation of SMAD protein import into nucleus negative regulation of SMAD protein import into nucleus detection of hypoxia endocardial cushion to mesenchymal transition vascular smooth muscle cell development positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation positive regulation of vascular smooth muscle cell differentiation atrioventricular canal morphogenesis regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis regulation of cell proliferation involved in heart morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline