Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • FLK1 (VEGF Receptor 2) Antibodies

          Invitrogen

          CD309 (FLK1) Monoclonal Antibody (Avas12a1), eBioscience™

          Advanced Verification
          48 Published Figures
          35 References
          View all (88) FLK1 (VEGF Receptor 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD309 (FLK1) Monoclonal Antibody (Avas12a1), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (48)
          • Advanced Verification (1)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 50

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD309 (FLK1) Antibody (14-5821-82) in Flow

          Surface staining of bEnd-3 cell line with Anti-Mouse CD309 (FLK1) PE.Appropriate isotype controls were used (open histogram).Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD309 (FLK1) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD309 (FLK1) Antibody in Immunocytochemistry (ICC/IF)
          CD309 (FLK1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD309 (FLK1) Antibody in Immunohistochemistry (IHC)
          CD309 (FLK1) Antibody in Immunohistochemistry (IHC)
          CD309 (FLK1) Antibody in Immunohistochemistry (IHC)
          CD309 (FLK1) Antibody in Immunohistochemistry (IHC)
          CD309 (FLK1) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD309 (FLK1) Antibody in Immunohistochemistry (IHC)
          CD309 (FLK1) Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry, Peptide array (Flow, ARRAY)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody

          Product Details

          14-5821-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          -

          Immunohistochemistry (IHC)

          Assay-Dependent
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 20 publications 20 publications

          Peptide Array (Array)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Dog, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Avas12a1

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • PE 
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467623

          Product Specific Information

          Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.

          Applications Reported: The Avas12a1 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining.

          Applications Tested: This Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD309; CD309 antigen; Fetal liver kinase 1; FLK-1; Kinase NYK; Protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR-2

          View more View less

          Gene Aliases: 6130401C07; Flk-1; Flk1; Kdr; Krd-1; Ly73; sVEGFR-2; VEGFR-2; VEGFR2

          View more View less

          UniProt ID: (Mouse) P35918

          View more View less

          Entrez Gene ID: (Mouse) 16542

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity integrin binding protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups growth factor binding vascular endothelial growth factor binding transmembrane signal receptor
          Process(es)
          angiogenesis ovarian follicle development vasculogenesis response to hypoxia positive regulation of protein phosphorylation regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation lymph vessel development cell migration involved in sprouting angiogenesis positive regulation of mesenchymal cell proliferation endocardium development endothelium development endochondral bone growth protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cytosolic calcium ion concentration multicellular organism development positive regulation of cell proliferation regulation of cell shape male gonad development positive regulation of endothelial cell migration positive regulation of phosphatidylinositol 3-kinase signaling phosphorylation cell migration peptidyl-tyrosine phosphorylation hemopoiesis cell differentiation lung development positive regulation of cell migration positive regulation of BMP signaling pathway positive regulation of vascular endothelial growth factor receptor signaling pathway positive regulation of TOR signaling embryonic hemopoiesis calcium-mediated signaling using intracellular calcium source cellular response to vascular endothelial growth factor stimulus peptidyl-tyrosine autophosphorylation vascular endothelial growth factor signaling pathway response to drug negative regulation of apoptotic process surfactant homeostasis positive regulation of MAPK cascade negative regulation of neuron apoptotic process cell fate commitment endothelial cell differentiation regulation of endothelial cell differentiation positive regulation of angiogenesis positive regulation of vasodilation protein autophosphorylation vascular endothelial growth factor receptor signaling pathway positive regulation of long-term neuronal synaptic plasticity lung alveolus development cell maturation branching morphogenesis of an epithelial tube neuron projection morphogenesis positive regulation of epithelial cell proliferation positive regulation of calcium-mediated signaling positive regulation of positive chemotaxis positive regulation of nitric-oxide synthase biosynthetic process positive regulation of focal adhesion assembly calcium ion homeostasis blood vessel endothelial cell differentiation positive regulation of ERK1 and ERK2 cascade regulation of hematopoietic progenitor cell differentiation negative regulation of endothelial cell apoptotic process positive regulation of vasculogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline