Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CXCL12 alpha Antibodies

          Invitrogen

          CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience™

          9 Published Figures
          16 References
          View all (7) CXCL12 alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (9)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CXCL12 alpha (SDF-1 alpha) Antibody (14-7992-81) in IHC

          Journal of cellular and molecular medicine 2020 - Tfh cells are an important source of CXCL12 in mice with schistosomiasis japonica. WT and icosl -/- mice were infected with 12 cercariae of Schistosoma japonicum per mouse, and sacrificed at 8 wk post-infection. (A, B) Spleens, mesenteric LN and livers from normal and infected mice were harvested, and cells were stained with CD3-percpcy5.5, CD4-PE-Cy7, CXCR5-FITC, PD-1-PE and CXCL12-APC antibodies. CXCL12 +... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          CXCL12 alpha (SDF-1 alpha) Antibody in Immunohistochemistry (IHC)
          CXCL12 alpha (SDF-1 alpha) Antibody in Western Blot, ELISA (WB, ELISA)
          CXCL12 alpha (SDF-1 alpha) Antibody in Flow Cytometry (Flow)

          Product Details

          14-7992-81

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 5 publications 5 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          1:300-1:500
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468513

          Product Specific Information

          Description: The polyclonal antibody reacts with both mouse and rat SDF-1 alpha. It was was generated using E. coli-expressed N terminal amio acids 20-89 of mouse SDF-1 alpha as an immunogen. SDF-1 alpha (stromal cell-derived factor-1 alpha), also known as pre-B-cell growth-stimulating factor (PBSF), is a 70-amino acid CXC chemokine originally cloned from a bone marrow stromal cell line. Targeted deletion of the SDF-1 gene resulted in defects of B-cell lymphopoiesis and bone marrow myelopoiesis. SDF-1 has been shown to be chemotactic for lymphocytes. In addition, SDF-1 was recently reported to be a ligand for CXCR4 (LESTR/fusin), a co-receptor for HIV-1 entry into T cells. SDF-1 binding to CXCR4 inhibits HIV-1 entry.

          Applications Reported: This polyclonal has been reported for use in immunoblotting (WB), immunoprecipitation, and immunohistochemistry. Cross-reactivity to SDF-1 alpha of other species has not been determined.

          Applications Tested: To achieve best results, titrate antibody by dilution for each application as follows:

          Western Blot 1:1000

          Immunoprecipitation 1:300-500

          Immunohistochemistry 1:100-500.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 12; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; CXCL; H-SDF-1-alpha; PBSF; Pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SDF 1 a; SDF 1 alpha; SDF 1α; SDF-1; SDF-1 gamma; SDF1 alpha; SDF1α; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; Thymic lymphoma cell-stimulating factor; TLSF; TPAR1

          View more View less

          Gene Aliases: Cxcl12; Pbsf; Scyb12; Sdf1; Tlsf; Tpar1

          View more View less

          UniProt ID: (Mouse) P40224

          View more View less

          Entrez Gene ID: (Rat) 24772, (Mouse) 20315

          View more View less

          Function(s)
          chemokine activity chemokine receptor binding CXCR chemokine receptor binding cytokine activity growth factor activity
          Process(es)
          patterning of blood vessels response to hypoxia ameboidal-type cell migration neuron migration positive regulation of endothelial cell proliferation immune response germ cell development brain development motor neuron axon guidance positive regulation of cell proliferation adult locomotory behavior germ cell migration response to radiation response to heat response to mechanical stimulus cell migration telencephalon cell migration regulation of cell migration positive regulation of cell migration organ regeneration positive regulation of dopamine secretion T cell proliferation response to peptide hormone positive regulation of neuron differentiation positive regulation of cell adhesion positive regulation of axon extension involved in axon guidance induction of positive chemotaxis regulation of calcium ion transport cell chemotaxis chemokine-mediated signaling pathway positive regulation of monocyte chemotaxis positive regulation of myoblast fusion negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of leukocyte tethering or rolling negative regulation of leukocyte apoptotic process positive regulation of T cell migration chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline