Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATSAVPSD NLPTYKLVVV GDGGVGKSAL TIQFFQKIFV PDYDPTIEDS YLKHTEIDNQ WAILDVLDTA G (1-70 aa encoded by BC047690)
MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein; MRAS; Muscle and microspikes Ras; Ras-related protein M-Ras; Ras-related protein R-Ras3; small GTP binding protein; unnamed protein product; X-Ras
Gene Aliases: 2900078C09Rik; AI326250; M-RAs; MRAS; NS11; R-RAS3; RRAS3; Xras
UniProt ID: (Human) O14807, (Mouse) O08989
Entrez Gene ID: (Human) 22808, (Mouse) 17532, (Rat) 25482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support