Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VRPKEKSVA GQVCLITGAG SGLGRLFALE FARRRALLVL WDINTQSNEE TAGMVRHIYR DLEAADAAAL QAGNGEEEIL PHCNLQVFTY TCDVGKRENV YLTAERVRKE VGEVSVLVNN AGVVSGHHLL ECPDELIERT MMVNCHAHFW TTKAFLPTML EINHGHIVTV ASSLGLFSTA GVEDYCASKF GVVGFHESLS HELKAAEKDG IKTTLVCPYL VDTGMFRGCR IRKEIEPFLP PLKPDYCVKQ AMKAILTDQP MICTPRLM (27-293 aa encoded by BC067131 )
RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Retinol dehydrogenase 10; retinol dehydrogenase 10 (all-trans); Short chain dehydrogenase/reductase family 16C member 4
Gene Aliases: 3110069K09Rik; 4921506A21Rik; AI875664; AW549993; D1Ertd762e; m366Asp; RDH10; SDR16C4; UNQ9375/PRO34191
UniProt ID: (Human) Q8IZV5, (Rat) Q80ZF7, (Mouse) Q8VCH7
Entrez Gene ID: (Human) 157506, (Rat) 353252, (Mouse) 98711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support