Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Syk Antibodies

          Proteintech

          SYK Polyclonal Antibody

          3 References
          View all (61) Syk antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SYK Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (16)
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SYK Antibody (14858-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human spleen tissue slide using 14858-1-AP (SYK antibody) at dilution of 1:200 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SYK Antibody in Western Blot (WB)
          SYK Antibody in Western Blot (WB)
          SYK Polyclonal Antibody

          Product Details

          14858-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,500
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SYK Fusion Protein Ag6657 (367-635 aa encoded by BC001645)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: RKLLTLEDK ELGSGNFGTV KKGYYQMKKV VKTVAVKILK NEANDPALKD ELLAEANVMQ QLDNPYIVRM IGICEAESWM LVMEMAELGP LNKYLQQNRH VKDKNIIELV HQVSMGMKYL EESNFVHRDL AARNVLLVTQ HYAKISDFGL SKALRADENY YKAQTHGKWP VKWYAPECIN YYKFSSKSDV WSFGVLMWEA FSYGQKPYRG MKGSEVTAML EKGERMGCPA GCPREMYDLM NLCWTYDVEN RPGFAAVELR LRNYYYDVVN (367-635 aa encoded by BC001645)

          Target Information

          Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72-Syk; p72Syk; SP1; Spleen tyrosine kinase; syk signaling; tyrosine-kinase syk; Tyrosine-protein kinase SYK

          View more View less

          Gene Aliases: p72-Syk; p72syk; ptk72; SYK; Sykb

          View more View less

          UniProt ID: (Human) P43405, (Rat) Q64725, (Mouse) P48025

          View more View less

          Entrez Gene ID: (Human) 6850, (Rat) 25155, (Mouse) 20963

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor signaling protein tyrosine kinase activity integrin binding protein binding ATP binding protein kinase binding receptor binding protein domain specific binding nucleotide binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups kinase modulator
          Process(es)
          angiogenesis lymph vessel development positive regulation of receptor internalization stimulatory C-type lectin receptor signaling pathway adaptive immune response macrophage activation involved in immune response neutrophil activation involved in immune response leukocyte activation involved in immune response serotonin secretion by platelet protein phosphorylation inflammatory response leukocyte cell-cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway integrin-mediated signaling pathway activation of JUN kinase activity cell proliferation organ morphogenesis regulation of platelet activation regulation of tumor necrosis factor-mediated signaling pathway viral process peptidyl-serine phosphorylation leukotriene biosynthetic process platelet activation neutrophil chemotaxis receptor internalization positive regulation of type I interferon production regulation of superoxide anion generation positive regulation of cell adhesion mediated by integrin peptidyl-tyrosine autophosphorylation Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis defense response to bacterium transcription factor import into nucleus positive regulation of mast cell degranulation regulation of neutrophil degranulation beta selection innate immune response positive regulation of interleukin-3 biosynthetic process positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process positive regulation of B cell differentiation positive regulation of gamma-delta T cell differentiation positive regulation of bone resorption positive regulation of alpha-beta T cell differentiation positive regulation of alpha-beta T cell proliferation blood vessel morphogenesis positive regulation of cytokine secretion positive regulation of peptidyl-tyrosine phosphorylation regulation of phagocytosis positive regulation of calcium-mediated signaling B cell receptor signaling pathway regulation of sequence-specific DNA binding transcription factor activity regulation of ERK1 and ERK2 cascade cellular response to molecule of fungal origin cellular response to low-density lipoprotein particle stimulus regulation of arachidonic acid secretion regulation of platelet aggregation activation of MAPK activity serotonin secretion cell surface receptor signaling pathway enzyme linked receptor protein signaling pathway cell migration peptidyl-tyrosine phosphorylation signal transduction by protein phosphorylation intracellular signal transduction protein autophosphorylation regulation of immune response immune system process G-protein coupled receptor signaling pathway phosphorylation cell differentiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0