Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
Immunogen sequence: EEIYKDRDS QITAIEKTFE DAQKSISQHY SKPRVTPVEV MPVFPDFKMW INPCAQVIFD SDPAPKDTSG AAALEMMSQA MIRGMMDEEG NQFVAYFLPV EETLKKRKRD QEEEMDYAPD DVYDYKIARE YNWNVKNKAS KGYEENYFFI FREGDGVYYN ELETRVRLSK RRAKAGVQSG TNALLVVKHR DMNEKELEAQ EARKAQLENH EPEEEEEEEM ETEEKEAGGS DEEQEKGSSS EKEGSEDEHS GSESEREEGD RDEASDKSGS GEDESSEDEA RAARDKEEIF GSDADSEDDA DSDDEDRGQA QGGSDNDSDS GSNGGGQRSR SHSRSASPFP SGSEHSAQED GSEAAASDSS EADSDSD (166-531 aa encoded by BC000017 )
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPAF1; Paf1, RNA polymerase II associated factor, homolog; Pancreatic differentiation protein 2; RNA polymerase II-associated factor 1 homolog; unnamed protein product
Gene Aliases: F23149_1; PAF1; PD2
UniProt ID: (Human) Q8N7H5
Entrez Gene ID: (Human) 54623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support