Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SLNSAHFDF QGCNFSEKNM YARDRRDGQS KEGSGRVAIY KASGIIHSYT LECNYNTGRS VNSIPAACHD NGRASPPPPP AFPSRYTVEL FEQVGRAMAI AALDMAECNP WPRIVLSEHS SLTNLRAWML KHVRNSRGLS STLNVGVNKK RGLRTPPKSH NGLPVSCSEN TLSRARSFST GTSAGGSSSS QQNSPQMKNS PSFPFHGSRP AGLPGLGSST QKVTHRVLGP VREPRSQDRR RQQQPLNHRP AGSLAPSPAP TSSGPASSHK LGSCLLPDSF NIPGSSCSLL SSGDKPEAVM VIGKGLLGTG ARMPCIKTRL QTCPRRVSAR RGPGFPRLGP GWAGAHRRLA EG (466-816 aa encoded by BC007415)
AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP/GTP binding protein like 5; ATP/GTP-binding protein-like 5; carboxypeptidase 5, cytosolic; cytosolic carboxypeptidase 5; Cytosolic carboxypeptidase-like protein 5; Protein deglutamylase CCP5; unnamed protein product
Gene Aliases: 4930455N08; 9430057O19Rik; AGBL5; CCP5; RP75
UniProt ID: (Human) A2VDI7, (Mouse) Q09M02
Entrez Gene ID: (Human) 60509, (Mouse) 231093
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support