Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAMSFEWPW QYRFPPFFTL QPNVDTRQKQ LAAWCSLVLS FCRLHKQSSM TVMEAQESPL FNNVKLQRKL PVESIQIVLE ELRKKGNLEW LDKSKSSFLI MWRRPEEWGK LIYQWVSRSG QNNSVFTLYE LTNGEDTEDE EFHGLDEATL LRALQALQQE HKAEIITVSD GRGVKFF (1-176 aa encoded by BC006282)
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dermal papilla-derived protein 9; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; HGNC:28122; hVps25; MGC10540; unnamed protein product; Vacuolar protein-sorting-associated protein 25
Gene Aliases: 1110020N13Rik; AW107467; D11Wsu68e; DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) Q9BRG1, (Rat) P0C0A1, (Mouse) Q9CQ80
Entrez Gene ID: (Human) 84313, (Rat) 681059, (Mouse) 28084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support