Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD3 Antibodies

          Invitrogen

          CD3e Monoclonal Antibody (145-2C11), Functional Grade, eBioscience™

          92 Published Figures
          368 References
          View all (474) CD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD3e Monoclonal Antibody (145-2C11), Functional Grade, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (92)
          CD3e Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD3e Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 93

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD3e Antibody (16-0031-82) in Flow

          Staining of mouse splenocytes with Anti-Mouse CD3e FITC (left) or PE (right). Appropriate isotype controls were used (open histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          CD3e Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunocytochemistry (ICC/IF)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD3e Antibody in Western Blot (WB)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry, T-Cell Activation (Flow, TCA)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in T-Cell Activation (TCA)
          CD3e Antibody in T-Cell Activation (TCA)
          CD3e Antibody in T-Cell Activation (TCA)
          CD3e Antibody in T-Cell Activation (TCA)
          CD3e Antibody in T-Cell Activation (TCA)
          CD3e Antibody in T-Cell Activation (TCA)

          Product Details

          16-0031-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 7 publications 7 publications

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 187 publications 187 publications

          ELISA (ELISA)

          -
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          Assay-Dependent
          View 104 publications 104 publications

          Inhibition Assays (Inhibition)

          -
          View 2 publications 2 publications

          T-Cell Activation (TCA)

          -
          View 16 publications 16 publications

          In vitro Assay (In vitro)

          -
          View 6 publications 6 publications

          Miscellaneous PubMed (Misc)

          -
          View 14 publications 14 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          145-2C11

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • NovaFluor Blue 510
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Red 685
          • NovaFluor Red 700
          • NovaFluor Red 710
          • NovaFluor Yellow 610
          • NovaFluor Yellow 660
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • NovaFluor Yellow 730
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468847

          Product Specific Information

          Description: The 145-2C11 monoclonal antibody reacts with mouse CD3e, a 20 kDa subunit of the TCR complex. Along with the other CD3 subunits, gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.

          Applications Reported:The 145-2C11 antibody has been reported for use in flow cytometric analysis. It has also been reported in cell activation and cell depletion. Please visit the BestProtocols® webpage to view a protocol for in vitro T-cell activation.

          Applications Tested: The 145-2C11 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level:Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation:Less than 10%, as determined by HPLC.

          Target Information

          The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD247; CD247 antigen; CD3 antigen, zeta polypeptide; CD3 TCR complex; CD3-zeta/eta; CD3d; CD3e; CD3g; FLJ18683; Leu-4; T cell antigen receptor complex epsilon subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T3/TCR complex

          View more View less

          Gene Aliases: 4930549J05Rik; A430104F18Rik; AI504783; AW552088; Cd247; CD3; Cd3-eta; Cd3-zeta; Cd3d; Cd3e; CD3epsilon; Cd3g; Cd3h; Cd3z; Cd3zeta; Ctg-3; Ctg3; T3d; T3e; T3g; T3z; Tcrk; Tcrz

          View more View less

          UniProt ID: (Mouse) P24161, (Mouse) P04235, (Mouse) P22646, (Mouse) P11942

          View more View less

          Entrez Gene ID: (Mouse) 12503, (Mouse) 12500, (Mouse) 12501, (Mouse) 12502

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding identical protein binding protein heterodimerization activity signal transducer activity SH3 domain binding defense/immunity protein immunoglobulin receptor superfamily
          Process(es)
          cell surface receptor signaling pathway T cell receptor signaling pathway protein complex assembly T cell activation positive thymic T cell selection positive regulation of T cell anergy response to nutrient T cell costimulation positive regulation of interferon-gamma production positive regulation of interleukin-4 production positive regulation of T cell proliferation negative thymic T cell selection positive regulation of interleukin-2 biosynthetic process negative regulation of smoothened signaling pathway positive regulation of alpha-beta T cell proliferation lymphocyte activation positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling positive regulation of T cell activation apoptotic signaling pathway establishment or maintenance of cell polarity protein transport regulation of lymphocyte apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline