Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • B220 (CD45R) Antibodies

          Invitrogen

          CD45R (B220) Monoclonal Antibody (RA3-6B2), Functional Grade, eBioscience™

          Advanced Verification
          93 Published Figures
          128 References
          View all (92) B220 (CD45R) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD45R (B220) Monoclonal Antibody (RA3-6B2), Functional Grade, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (93)
          • Advanced Verification (1)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 95

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD45R (B220) Antibody (16-0452-82) in Flow

          Staining of mouse splenocytes with Anti-Human/Mouse CD45R (B220) FITC (left) or APC (right).Appropriate isotype controls were used (open histogram).Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD45R (B220) Antibody in Immunocytochemistry (ICC/IF)
          CD45R (B220) Antibody in Immunocytochemistry (ICC/IF)
          CD45R (B220) Antibody in Immunocytochemistry (ICC/IF)
          CD45R (B220) Antibody in Immunocytochemistry (ICC/IF)
          CD45R (B220) Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          CD45R (B220) Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          CD45R (B220) Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Immunohistochemistry (IHC)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody in Flow Cytometry (Flow)
          CD45R (B220) Antibody

          Product Details

          16-0452-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 36 publications 36 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 79 publications 79 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Fish, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RA3-6B2

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • eFluor 570
          • eFluor 615
          • eFluor 660
          • FITC
          • NovaFluor Blue 510
          • NovaFluor Blue 530
          • NovaFluor Blue 585
          • NovaFluor Blue 610-30S
          • NovaFluor Blue 660-40S
          • NovaFluor Blue 725
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 700
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 660
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468959

          Product Specific Information

          Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.

          Applications Reported: The RA3-6B2 antibody has been reported for use in flow cytometric analysis. RA3-6B2 has also been reported in functional studies of B cells.

          Applications Tested: The RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD45; CD45 antigen; glycoprotein 180; L-CA; Leukocyte common antigen; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; protein tyrosine phosphatase; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen; unnamed protein product

          View more View less

          Gene Aliases: B220; CD45; CD45R; GP180; IMD105; L-CA; LCA; loc; Ly-5; LY5; Lyt-4; PTPRC; T200

          View more View less

          UniProt ID: (Human) P08575, (Mouse) P06800

          View more View less

          Entrez Gene ID: (Human) 5788, (Mouse) 19264

          View more View less

          Function(s)
          phosphoprotein phosphatase activity protein tyrosine phosphatase activity transmembrane receptor protein tyrosine phosphatase activity receptor binding protein binding heparin binding hydrolase activity protein kinase binding protein tyrosine kinase inhibitor activity ankyrin binding spectrin binding heparan sulfate proteoglycan binding phosphatase activity protein phosphatase protein modifying enzyme
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade natural killer cell differentiation negative regulation of T cell mediated cytotoxicity positive regulation of T cell mediated cytotoxicity negative regulation of cytokine-mediated signaling pathway hematopoietic progenitor cell differentiation positive regulation of immunoglobulin production positive regulation of T cell mediated immunity regulation of humoral immune response mediated by circulating immunoglobulin positive regulation of humoral immune response mediated by circulating immunoglobulin negative regulation of protein kinase activity protein dephosphorylation negative regulation of cell adhesion involved in substrate-bound cell migration leukocyte cell-cell adhesion signal transduction cell surface receptor signaling pathway response to gamma radiation regulation of gene expression dephosphorylation B cell differentiation T cell differentiation positive regulation of B cell proliferation regulation of interleukin-8 production negative regulation of interleukin-2 production positive regulation of interleukin-2 production positive regulation of tumor necrosis factor production heterotypic cell-cell adhesion T cell proliferation B cell proliferation positive regulation of T cell proliferation T cell activation gamma-delta T cell differentiation positive regulation of MAPK cascade cell cycle phase transition plasma membrane raft distribution positive thymic T cell selection negative thymic T cell selection positive regulation of T cell differentiation positive regulation of gamma-delta T cell differentiation positive regulation of protein kinase activity negative regulation of JAK-STAT cascade alpha-beta T cell proliferation positive regulation of alpha-beta T cell proliferation positive regulation of isotype switching to IgG isotypes bone marrow development stem cell development regulation of phagocytosis positive regulation of phagocytosis positive regulation of calcium-mediated signaling T cell receptor signaling pathway B cell receptor signaling pathway regulation of T cell receptor signaling pathway positive regulation of antigen receptor-mediated signaling pathway release of sequestered calcium ion into cytosol defense response to virus regulation of cell cycle positive regulation of Fc receptor mediated stimulatory signaling pathway negative regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade extrinsic apoptotic signaling pathway negative regulation of interleukin-4-mediated signaling pathway negative regulation of microglial cell activation response to aldosterone DN2 thymocyte differentiation positive regulation of hematopoietic stem cell migration positive regulation of stem cell proliferation regulation of extrinsic apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway activation of MAPK activity immunoglobulin biosynthetic process negative regulation of protein autophosphorylation peptidyl-tyrosine dephosphorylation regulation of B cell differentiation negative regulation of peptidyl-tyrosine phosphorylation regulation of B cell receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline