Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QLLKRLGSG QFGEVWEGLW NNTTPVAVKT LKPGSMDPND FLREAQIMKN LRHPKLIQLY AVCTLEDPIY IITELMRHGS LQEYLQNDTG SKIHLTQQVD MAAQVASGMA YLESRNYIHR DLAARNVLVG EHNIYKVADF GLARVFKVDN EDIYESRHEI KLPVKWTAPE AIRSNKFSIK SDVWSFGILL YEIITYGKMP YSGMTGAQVI QMLAQNYRLP QPSNCPQQFY NIMLECWNAE PKERPTFETL RWKLEDYFET DSSYSDANNF IR (235-505 aa encoded by BC012916)
FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell src-homology tyrosine kinase; Beta-cell Src-homology tyrosine kinase; BSK; EC 2.7.1.112; EC 2.7.10.2; FYN-related kinase; GASK; Gastrointestinal-associated kinase; Gastrointestinal-associated tyrosine kinase; GTK; Intestine tyrosine kinase; Nuclear tyrosine protein kinase RAK; Protein-tyrosine kinase 5; PTK5 protein tyrosine kinase 5; src related tyrosine kinase; Tyrosine-protein kinase FRK; unnamed protein product
Gene Aliases: BSK; BSK/IYK; C85044; FRK; Gask; GTK; Iyk; PTK5; RAK
UniProt ID: (Human) P42685, (Mouse) Q922K9, (Rat) Q62662
Entrez Gene ID: (Human) 2444, (Mouse) 14302, (Rat) 79209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support