Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PCSK5 Antibodies

          Proteintech

          PC6 Polyclonal Antibody

          2 References
          View all (4) PCSK5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PC6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (19)
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PC6 Antibody (16470-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 16470-1-AP) (PCSK5 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Western Blot (WB)
          PC6 Polyclonal Antibody

          Product Details

          16470-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PC6 Fusion Protein Ag9552 (32-351 aa encoded by BC012064)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.18 mg/mL

          Amount

          27 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TRVYTNHWA VKIAGGFPEA NRIASKYGFI NIGQIGALKD YYHFYHSRTI KRSVISSRGT HSFISMEPKV EWIQQQVVKK RTKRDYDFSR AQSTYFNDPK WPSMWYMHCS DNTHPCQSDM NIEGAWKRGY TGKNIVVTIL DDGIERTHPD LMQNYDALAS CDVNGNDLDP MPRYDASNEN KHGTRCAGEV AAAANNSHCT VGIAFNAKIG GVRMLDGDVT DMVEAKSVSF NPQHVHIYSA SWGPDDDGKT VDGPAPLTRQ AFENGVRMGR RGLGSVFVWA SGNGGRSKDH CSCDGYTNSI YTISISSTAE SGKKPWYLEE C (32-351 aa encoded by BC012064)

          Target Information

          PCSK5 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. PCSK5 mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: PC5; PC6; prohormone convertase 5; Proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; Proprotein convertase subtilisin/kexin type 5; protease PC6; SPC6; subtilase; Subtilisin-like proprotein convertase 6; Subtilisin/kexin-like protease PC5; unnamed protein product

          View more View less

          Gene Aliases: b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; SPC6

          View more View less

          UniProt ID: (Human) Q92824, (Mouse) Q04592

          View more View less

          Entrez Gene ID: (Human) 5125, (Mouse) 18552

          View more View less

          Function(s)
          endopeptidase activity serine-type endopeptidase activity protein binding peptidase activity serine-type peptidase activity hydrolase activity peptide binding serine protease
          Process(es)
          kidney development renin secretion into blood stream signal peptide processing proteolysis cell-cell signaling heart development female pregnancy embryo implantation anterior/posterior pattern specification protein processing peptide hormone processing viral life cycle respiratory tube development secretion by cell plasma lipoprotein particle remodeling limb morphogenesis peptide biosynthetic process embryonic digestive tract development embryonic skeletal system development cytokine precursor processing cytokine biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline