Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAAALLARA SGPARRALCP RAWRQLHTIY QSVELPETHQ MLLQTCRDFA EKELFPIAAQ VDKEHLFPAA QVKKMGGLGL LAMDVPEELG GAGLDYLAYA IAMEEISRGC ASTGVIMSVN NSLYLGPILK FGSKEQKQAW VTPFTSGDKI GCFALSEPGN GSDAGAASTT ARAEGDSWVL NGTKAWITNA WEASAAVVFA STDRALQNKS ISAFLVPMPT PGLTLGKKED KLGIRGSSTA NLIFEDCRIP KDSILGEPGM GFKIAMQTLD MGRIGIASQA LGIAQTALDC AVNYAENRMA FGAPLTKLQV IQFKLADMAL ALESARLLTW RAAMLKDNKK PFIKEAAMAK LAASEAATAI SHQAIQILGG MGYVTEMPAE RHYRDARITE IYEGTSEIQR LVIAGHLLRS YRS (1-412 aa encoded by BC025963)
Introduces a double bond at position 2 in saturated acyl-CoA's of short chain length, i. e. less than 6 carbon atoms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetyl-Coenzyme A dehydrogenase, short chain; acyl-CoA dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, short chain; Butyryl-CoA dehydrogenase; epididymis secretory sperm binding protein; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; short chain acyl-CoA dehydrogenase precursor (EC 1.3.99.2); short chain acyl-coenzyme A dehydrogenase; short-chain acyl-CoA dehydrogenase; Short-chain specific acyl-CoA dehydrogenase, mitochondrial; unnamed protein product; unsaturated acyl-CoA reductase
Gene Aliases: ACAD3; ACADS; AI196007; Bcd-1; Bcd1; Hdlq8; SCAD
UniProt ID: (Human) P16219, (Rat) P15651, (Mouse) Q07417
Entrez Gene ID: (Human) 35, (Rat) 64304, (Mouse) 11409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support