Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • CSF1R (CD115) Antibodies

          Invitrogen

          CD115 (c-fms) Monoclonal Antibody (AFS98), APC, eBioscience™

          28 Published Figures
          103 References
          View all (85) CSF1R (CD115) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD115 (c-fms) Monoclonal Antibody (AFS98), APC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (28)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 29

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD115 (c-fms) Antibody (17-1152-82) in Flow

          Staining of thioglycolate-induced peritoneal exudate cells (PECs) with 0.03 µg of Rat IgG2a kappa Isotype Control APC (Product # 17-4321-81) (blue histogram) or 0.03 µg of Anti-Mouse CD115 (c-fms) APC (purple histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)
          CD115 (c-fms) Antibody in Flow Cytometry (Flow)

          Product Details

          17-1152-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.06 µg/test
          View 103 publications 103 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Fish, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AFS98

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 488 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 563
          • Functional Grade
          • NovaFluor Red 685
          • NovaFluor Red 710
          • NovaFluor Yellow 610
          • NovaFluor Yellow 730
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1210789

          Product Specific Information

          Description: The AFS98 monoclonal antibody reacts with the mouse CD115 molecule, a receptor for macrophage colony stimulating factor (M-CSF) or colony stimulating factor-1 (CSF-1). CD115 is expressed by monocyte, macrophage, osteoclast, and some epithelial cells. It is a 150 kDa c-fms gene product and belongs to immunoglobulin family. CSF-1 signaling through CSF-1R regulates the proliferation and differentiation of cells in the monocytic lineage.

          Applications Reported: This AFS98 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This AFS98 antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: c-fms; CD115; CSF-1 receptor; CSF-1-R; CSFIR; EC 2.7.10.1; kinase CSFR; Macrophage colony-stimulating factor 1 receptor; OTTHUMP00000224093; Proto-oncogene c-Fms; proto-oncogene fms

          View more View less

          Gene Aliases: AI323359; CD115; CSF-1R; Csf1r; Csfmr; Fim-2; Fms; M-CSF-R; M-CSFR

          View more View less

          UniProt ID: (Mouse) P09581

          View more View less

          Entrez Gene ID: (Mouse) 12978

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity macrophage colony-stimulating factor receptor activity protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein phosphatase binding cytokine binding protein homodimerization activity transmembrane signal receptor
          Process(es)
          positive regulation of protein phosphorylation immune system process protein phosphorylation inflammatory response transmembrane receptor protein tyrosine kinase signaling pathway cell proliferation positive regulation of cell proliferation regulation of cell shape phosphorylation peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway hemopoiesis osteoclast differentiation positive regulation of cell migration ruffle organization cellular response to macrophage colony-stimulating factor stimulus macrophage colony-stimulating factor signaling pathway positive regulation of tyrosine phosphorylation of Stat3 protein innate immune response regulation of bone resorption cell-cell junction maintenance positive regulation of osteoclast differentiation phosphatidylinositol metabolic process protein autophosphorylation phosphatidylinositol-mediated signaling positive regulation of protein tyrosine kinase activity positive regulation of ERK1 and ERK2 cascade cellular response to cytokine stimulus positive regulation of protein serine/threonine kinase activity positive regulation of chemokine secretion positive regulation of cell motility regulation of actin cytoskeleton reorganization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline