Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • c-Kit (CD117) Antibodies

          Invitrogen

          CD117 (c-Kit) Monoclonal Antibody (2B8), APC, eBioscience™

          53 Published Figures
          257 References
          View all (227) c-Kit (CD117) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD117 (c-Kit) Monoclonal Antibody (2B8), APC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (53)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 54

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD117 (c-Kit) Antibody (17-1171-82) in Flow

          Staining of C57Bl/6 bone marrow cells with Mouse Hematopoietic Lineage eFluor® 450 Cocktail (Product # 88-7772-72), Anti-Mouse Ly-6A/E (Sca-1) PerCP-Cyanine5-5 (Product # 45-5981-82) and 0.06 µg of Rat IgG2b K Isotype Control APC (Product # 17-4031-82) (left) or 0.06 µg of Anti-Mouse CD117 (c-Kit) APC (right). Lineage negative/low, viable cells, as determined by Fixable Viability Dye eFluor® 780 (Product # 65-0865-14), were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)

          Product Details

          17-1171-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 246 publications 246 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Affinity Purification (AP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Mouse, Pig

          Published species

          Fish, Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2B8

          Conjugate

          APC APC APC
          • Unconjugated
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_469430

          Product Specific Information

          Description: The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145-kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells.

          Applications Reported: The 2B8 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The 2B8 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: belly-spot; c-kit; c-kit proto-oncogene protein; CD117; ckit; dominant spotting; Dominant white spotting; Mast/stem cell growth factor receptor Kit; Proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; SCFR; spotted sterile male; Steel Factor Receptor; Tyrosine-protein kinase Kit

          View more View less

          Gene Aliases: Bs; c-KIT; CD117; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; MGF; SCO1; SCO5; Sl; SOW3; Ssm; Tr-kit; W

          View more View less

          UniProt ID: (Pig) Q2HWD6, (Mouse) P05532

          View more View less

          Entrez Gene ID: (Pig) 396810, (Mouse) 16590

          View more View less

          Function(s)
          transmembrane receptor protein tyrosine kinase activity ATP binding cytokine binding metal ion binding nucleotide binding protease binding protein kinase activity protein tyrosine kinase activity receptor signaling protein tyrosine kinase activity stem cell factor receptor activity protein binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein homodimerization activity transmembrane signal receptor
          Process(es)
          activation of MAPK activity ovarian follicle development immature B cell differentiation dendritic cell cytokine production inflammatory response spermatogenesis regulation of cell shape peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway lamellipodium assembly T cell differentiation erythrocyte differentiation melanocyte differentiation actin cytoskeleton reorganization mast cell cytokine production embryonic hemopoiesis megakaryocyte development Fc receptor signaling pathway Kit signaling pathway erythropoietin-mediated signaling pathway mast cell degranulation pigmentation digestive tract development stem cell differentiation detection of mechanical stimulus involved in sensory perception of sound cell chemotaxis mast cell differentiation melanocyte migration melanocyte adhesion myeloid progenitor cell differentiation lymphoid progenitor cell differentiation mast cell chemotaxis myeloid leukocyte differentiation protein phosphorylation glycosphingolipid metabolic process chemotaxis transmembrane receptor protein tyrosine kinase signaling pathway germ cell development spermatid development positive regulation of cell proliferation germ cell migration visual learning response to radiation positive regulation of gene expression phosphorylation hemopoiesis cell differentiation positive regulation of cell migration positive regulation of pseudopodium assembly somatic stem cell population maintenance ectopic germ cell programmed cell death intracellular signal transduction positive regulation of tyrosine phosphorylation of Stat1 protein positive regulation of tyrosine phosphorylation of Stat3 protein positive regulation of tyrosine phosphorylation of Stat5 protein negative regulation of programmed cell death positive regulation of MAP kinase activity positive regulation of MAPK cascade positive regulation of Notch signaling pathway positive regulation of JAK-STAT cascade protein autophosphorylation developmental pigmentation regulation of developmental pigmentation somatic stem cell division positive regulation of long-term neuronal synaptic plasticity epithelial cell proliferation positive regulation of sequence-specific DNA binding transcription factor activity cellular response to thyroid hormone stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline