Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RAB27A Antibodies

          Proteintech

          RAB27A Polyclonal Antibody

          53 References
          View all (15) RAB27A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAB27A Polyclonal Antibody

          • Antibody Testing Data (16)
          RAB27A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB27A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB27A Antibody (17817-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using RAB27A antibody 17817-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB27A Antibody in Immunocytochemistry (ICC/IF)
          RAB27A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Western Blot (WB)
          RAB27A Antibody in Immunoprecipitation (IP)
          RAB27A Polyclonal Antibody

          Product Details

          17817-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 44 publications 44 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Chicken, Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAB27A Fusion Protein Ag12136 (1-221 aa encoded by BC107680)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSDGDYDYL IKFLALGDSG VGKTSVLYQY TDGKFNSKFI TTVGIDFREK RVVYRASGPD GATGRGQRIH LQLWDTAGQE RFRSLTTAFF RDAMGFLLLF DLTNEQSFLN VRNWISQLQM HAYCENPDIV LCGNKSDLED QRVVKEEEAI ALAEKYGIPY FETSAANGTN ISQAIEMLLD LIMKRMERCV DKSWIPEGVV RSNGHASTDQ LSEEKEKGAC GC (1-221 aa encoded by BC107680)

          Target Information

          The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GTP-binding protein Ram; mutant Ras-related protein Rab-27A; Rab-27; Ras-related protein Rab-27A

          View more View less

          Gene Aliases: GS2; HsT18676; RAB27; RAB27A; RAM

          View more View less

          UniProt ID: (Human) P51159

          View more View less

          Entrez Gene ID: (Human) 5873

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding GDP binding protein domain specific binding myosin V binding
          Process(es)
          protein targeting exocytosis small GTPase mediated signal transduction blood coagulation positive regulation of gene expression antigen processing and presentation melanocyte differentiation melanosome localization melanosome transport multivesicular body organization cytotoxic T cell degranulation natural killer cell degranulation positive regulation of exocytosis synaptic vesicle transport positive regulation of phagocytosis multivesicular body sorting pathway complement-dependent cytotoxicity positive regulation of regulated secretory pathway positive regulation of reactive oxygen species biosynthetic process positive regulation of constitutive secretory pathway exosomal secretion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gbgwf:80/100.66.79.18:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0