Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEFNPSDHP RASTIFLSKS QTDVREKRKS LFINHHPPGQ IARKYSSCST IFLDDSTVSQ PNLKYTIKCV ALAIYYHIKN RDPDGRMLLD IFDENLHPLS KSEVPPDYDK HNPEQKQIYR FVRTLFSAAQ LTAECAIVTL VYLERLLTYA EIDICPANWK RIVLGAILLA SKVWDDQAVW NVDYCQILKD ITVEDMNELE RQFLELLQFN INVPSSVYAK YYFDLRSLAE ANNLSFPLEP LSRERAHKLE AISRLCEDKY KDLRRSARKR SASADNLTLP RWSPAIIS (1-287 aa encoded by BC104773)
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBCP1; Cyc-Y; Cyclin box protein 1; Cyclin fold protein 1; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; unnamed protein product
Gene Aliases: 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; CCNY; CFP1; RGD1565969
UniProt ID: (Human) Q8ND76, (Mouse) Q8BGU5
Entrez Gene ID: (Human) 219771, (Mouse) 67974, (Rat) 361261
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support