Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MAD1 Antibodies

          Proteintech

          MAD1 Polyclonal Antibody

          5 References
          View all (16) MAD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MAD1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (24)
          MAD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MAD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAD1 Antibody (18322-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using MAD1 antibody (Product # 18322-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAD1 Antibody in Immunocytochemistry (ICC/IF)
          MAD1 Antibody in Immunocytochemistry (ICC/IF)
          MAD1 Antibody in Immunocytochemistry (ICC/IF)
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Western Blot (WB)
          MAD1 Antibody in Immunoprecipitation (IP)
          MAD1 Polyclonal Antibody

          Product Details

          18322-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAD1 Fusion Protein Ag13171 (370-718 aa encoded by BC009964)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: QEELRQVSG QLLEERKKRE THEALARRLQ KRVLLLTKER DGMRAILGSY DSELTPAEYS PQLTRRMREA EDMVQKVHSH SAEMEAQLSQ ALEELGGQKQ RADMLEMELK MLKSQSSSAE QSFLFSREEA DTLRLKVEEL EGERSRLEEE KRMLEAQLER RALQGDYDQS RTKVLHMSLN PTSVARQRLR EDHSQLQAEC ERLRGLLRAM ERGGTVPADL EAAAASLPSS KEVAELKKQV ESAELKNQRL KEVFQTKIQE FRKACYTLTG YQIDITTENQ YRLTSLYAEH PGDCLIFKAT SPSGSKMQLL ETEFSHTVGE LIEVHLRRQD SIPAFLSSLT LELFSRQTVA (370-718 aa encoded by BC009964)

          Target Information

          MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hMAD1; HsMAD1; MAD1 mitotic arrest deficient like 1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; Mitotic arrest deficient 1-like protein 1; Mitotic checkpoint MAD1 protein homolog; Mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; Tax-binding protein 181; tumor protein p53 inducible protein 9; unnamed protein product

          View more View less

          Gene Aliases: AI173502; AW550425; MAD1; MAD1L1; MVA7; PIG9; TP53I9; TXBP181

          View more View less

          UniProt ID: (Human) Q9Y6D9, (Mouse) Q9WTX8

          View more View less

          Entrez Gene ID: (Human) 8379, (Rat) 680006, (Mouse) 17120

          View more View less

          Function(s)
          protein binding identical protein binding kinetochore binding
          Process(es)
          mitotic spindle assembly checkpoint negative regulation of T cell proliferation thymus development regulation of metaphase plate congression regulation of mitotic cell cycle phase transition cell cycle mitotic nuclear division cell division attachment of mitotic spindle microtubules to kinetochore positive regulation of mitotic cell cycle spindle assembly checkpoint negative regulation of mitotic spindle assembly checkpoint
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline