Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EAVYCKFHY KTDQGIKNLS VEDAARLSQE DPDYGIRDLF NAIATGKYPS WTFYIQVMTF NQAETFPFNP FDLTKVWPHK DYPLIPVGKL VLNRNPVNYF AEVEQIAFDP SNMPPGIEAS PDKMLQGRLF AYPDTHRHRL GPNYLHIPVN CPYRARVANY QRDGPMCMQD NQGGAPNYYP NSFGAPEQQP SALEHSIQYS GEVRRFNTAN DDNVTQVRAF YVNVLNEEQR KRLCENIAGH LKDAQIFIQK KAVKNFTEVH PDYGSHIQAL LDKYNAEKPK NAIHTFVQSG SHLAAREKAN L (228-527 aa encoded by BC112219)
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAT; Catalase; catalase 1; CTA1; epididymis secretory sperm binding protein; HGNC:1516; MGC138422; MGC138424; unnamed protein product
Gene Aliases: 2210418N07; Cas-1; Cas1; CAT; Cat01; Catl; Cs-1; CS1
UniProt ID: (Human) P04040, (Rat) P04762, (Mouse) P24270
Entrez Gene ID: (Human) 847, (Rat) 24248, (Mouse) 12359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support