Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHRNA7 Antibodies

          Proteintech

          CHRNA7 Polyclonal Antibody

          18 References
          View all (19) CHRNA7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CHRNA7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CHRNA7 Antibody (21379-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded Mouse brain tissue slide using (Product # 21379-1-AP) CHRNA7 antibody at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Western Blot (WB)
          CHRNA7 Antibody in Flow Cytometry (Flow)
          CHRNA7 Polyclonal Antibody

          Product Details

          21379-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 12 publications 12 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CHRNA7 Fusion Protein Ag15980 (130-321 aa encoded by BC037571)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TVIVLQYHH HDPDGGKMPK WTRVILLNWC AWFLRMKRPG EDKVRPACQH KQRRCSLASV EMSAVAPPPA SNGNLLYIGF RGLDGVHCVP TPDSGVVCGR MACSPTHDEH LLHGGQPPEG DPDLAKILEE VRYIANRFRC QDESEAVCSE WKFAACVVDR LCLMAFSVFT IICTIGILMS APNFVEAVSK DFA (130-321 aa encoded by BC037571)

          Target Information

          Chromosome 15 encodes over 700 genes and comprises nearly 3% of the human genome. CHRFAM7A, a fusion protein comprised of FAM7A and AChR alpha7, is encoded by the CHRFAM7A gene, which is located on a region of human chromosome 15 that is associated with several neuropsychiatric disorders, such as schizophrenia and bipolar affective disorder. CHRFAM7A, also known as D-10 or CHRNA7, is a 412 amino acid multi-pass membrane protein that belongs to the nicotinic acetylcholine receptor (nAChR) subfamily of ligand-gated ion channel proteins, a protein superfamily that mediates signal transmission at synapses. CHRFAM7A is expressed in the hippocampus. Alternative splicing of the CHRFAM7A gene produces two splice variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: a7 nicotinic acetylcholine receptor; acetylcholine receptor; acetylcholine receptor alpha 7 neural; alpha 7 neuronal nicotinic acetylcholine receptor; alpha-7 nicotinic cholinergic receptor subunit; alpha7 nicotinic receptor; alpha7-nAChR; bungarotoxin alpha; C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); C holinergic receptor, nicotinic, alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); cholinergic receptor; cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; CHRFAM7A; CHRNA7-DR1; cytotransmitter gated ion channel; D-10; nAChR alpha7; nAChR7; neuronal acetylcholine receptor protein, alpha-7 chain; Neuronal acetylcholine receptor subunit alpha-7; neuronal nicotinic acetycholine receptor alpha 7; Nicotinic acetylcholine receptor subunit alpha-7; nicotinic receptor alpha 7 subunit; unnamed protein product

          View more View less

          Gene Aliases: Acra7; alpha7; BTX; CHRNA7; CHRNA7-2; nAchR; nAChR7; NACHRA7; NARAD

          View more View less

          UniProt ID: (Human) P36544, (Rat) Q05941, (Mouse) P49582

          View more View less

          Entrez Gene ID: (Human) 1139, (Rat) 25302, (Mouse) 11441

          View more View less

          Function(s)
          beta-amyloid binding transmembrane signaling receptor activity ion channel activity extracellular ligand-gated ion channel activity calcium channel activity protein binding acetylcholine receptor activity toxic substance binding chloride channel regulator activity acetylcholine-gated cation channel activity acetylcholine binding protein homodimerization activity acetylcholine-activated cation-selective channel activity drug binding adenylate cyclase binding protein kinase binding ATPase binding scaffold protein binding ligand-gated ion channel
          Process(es)
          response to hypoxia ion transport calcium ion transport cellular calcium ion homeostasis signal transduction I-kappaB kinase/NF-kappaB signaling chemical synaptic transmission synaptic transmission, cholinergic learning or memory memory short-term memory positive regulation of cell proliferation signal transduction involved in regulation of gene expression negative regulation of tumor necrosis factor production ion transmembrane transport response to nicotine regulation of membrane potential negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAPK cascade positive regulation of angiogenesis synapse organization cognition sensory processing positive regulation of protein metabolic process excitatory postsynaptic potential synaptic transmission involved in micturition positive regulation of ERK1 and ERK2 cascade calcium ion transmembrane transport acetylcholine receptor signaling pathway dendritic spine organization modulation of excitatory postsynaptic potential dendrite arborization negative regulation of cytokine production involved in inflammatory response positive regulation of long-term synaptic potentiation positive regulation of beta-amyloid formation negative regulation of beta-amyloid formation regulation of amyloid precursor protein catabolic process response to amyloid-beta response to acetylcholine negative regulation of 3'-UTR-mediated mRNA stabilization regulation of amyloid fibril formation ligand-gated ion channel signaling pathway positive regulation of excitatory postsynaptic potential activation of MAPK activity positive regulation of heart rate involved in baroreceptor response to decreased systemic arterial blood pressure endocytosis associative learning regulation of norepinephrine secretion neuronal action potential sperm motility response to food regulation of synaptic transmission, dopaminergic negative regulation of interleukin-1 beta production negative regulation of interleukin-6 production behavioral response to nicotine T cell activation B cell activation dopamine biosynthetic process ovulation cycle response to ethanol behavioral response to ethanol regulation of inflammatory response negative regulation of inflammatory response generation of ovulation cycle rhythm cation transmembrane transport transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline