Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IDGGAGGNG LVGPGGSGAG PGGGLTPSAP PYGAAFPPFP EGHPAVLPGE DPPPYSPLTS PDSGSAPMIT (15-83 aa encoded by BC002867)
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 4,5-bisphosphate. Does not hydrolyze phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-bisphosphate, inositol 3,5-bisphosphate, inositol 3,4-bisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4-monophosphate and phosphatidylinositol 3-monophosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PtdIns-4,5-P(2) 4-phosphatase type I; PtdIns-4,5-P2 4-Ptase I; Transmembrane protein 55B; transmembrane protein 55B-like; Type 1 phosphatidylinositol 4,5-bisphosphate 4-phosphatase; Type 1 PtdIns-4,5-P2 4-Ptase; type I phosphatidylinositol 4,5-bisphosphate 4-phosphatase; type I phosphatidylinositol-4,5-bisphosphate 4-phosphatase; type I PtdIns-4,5-P(2) 4-phosphatase; unnamed protein product
Gene Aliases: BC039161; C14orf9; PIP4P1; RGD1307475; TMEM55B
UniProt ID: (Human) Q86T03, (Rat) Q5PPM8, (Mouse) Q3TWL2
Entrez Gene ID: (Human) 90809, (Rat) 364298, (Mouse) 219024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support