Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSYAEKPDE ITKDEWMEKL NNLHVQRADM NRLIMNYLVT EGFKEAAEKF RMESGIEPSL DLETLDERIK IREMILKGQI QEAIALINSL HPELLDTNRY LYFHLQQQHL IELIRQRETE AALEFAQTQL AEQGEESREC LTEMERTLAL LAFDSPEESP FGDLLHTMQR QKVWSEVNQA VLDYENREST PKLAKLLKLL LWAQNELDQK KVKYPKMTDL SKGVIEEPK (1-228 aa encoded by BC032120 )
TWA1 was identified through a two hybrid-associated protein screen with RanBPM. TWA1 is well conserved through evolution and is localized within the nucleus. It interacts with RanBP9 and comprises a protein complex with RanBPM and Muskelin. TWA1 was found to possess the LisH-CTLH motif which is detected in proteins involved in microtubule dynamics, cell migration, nucleokinesis and chromosome segregation. These functions overlap with functions suggested for the RanGTPase cycle. Recent findings suggested that there is an as yet uncovered function of the RanGTPase cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BWK-1; Bwk1 leukemia-related; GID complex subunit 8 homolog; Glucose-induced degradation protein 8 homolog; protein C20orf11; protein C20orf11 homolog; Twa1; two hybrid associated protein 1; two hybrid associated protein No. 1 with RanBPM; Two hybrid-associated protein 1 with RanBPM; unnamed protein product
Gene Aliases: 2310003C23Rik; 4833420G11Rik; AI451474; Bwk1; C20orf11; GID8; TWA1
UniProt ID: (Human) Q9NWU2, (Mouse) Q9D7M1
Entrez Gene ID: (Human) 54994, (Mouse) 76425, (Rat) 296466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support