Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SCITSYTHR DYMVSNPSLI TKQMLTFYSA DTDQGQELTR LWDRVMIEQE CCGTSGPMDW VNFTSAFRAA TPEVVFPWPP LCCRRTGNFI PLNEEGCRLG HMDYLFTKGC FEHIGHAIDS YT (110-230 aa encoded by BC107098)
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetraspanin-21; Tspan-21; UP1a; UPIa; UPKa; Uroplakin Ia; Uroplakin-1a
Gene Aliases: 1110031P12Rik; AU019298; C87192; TSPAN21; UP1A; UPIA; Upk1; UPK1A; UPKA
UniProt ID: (Human) O00322, (Mouse) Q9D132
Entrez Gene ID: (Human) 11045, (Rat) 365227, (Mouse) 109637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support