Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RSPEASPKH SHSPASSPKP LKKQNPSSGA RLNQDKRSVA DEQVTAIVNK PLMSKAPTKE AGAVVPQSKY SGRHHIPNPS NGELHSQYHG YYVKLNAPQH PPVDVEDGDG SSQSSSALVH KPSANKWSPS KSVTKPVAKE SKAEPKAKKS ELAIPKNPAS NDSCPALEKE ANSGPNS (464-639 aa encoded by BC140875 )
Junctional complexes between the plasma membrane and endoplasmic/sarcoplasmic reticulum are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. The protein encoded by this gene is a component of junctional complexes and is composed of a C-terminal hydrophobic segment spanning the endoplasmic/sarcoplasmic reticulum membrane and a remaining cytoplasmic domain that shows specific affinity for the plasma membrane. This gene is a member of the junctophilin gene family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: JP-1; Junctophilin type 1; junctophilin type1; Junctophilin-1; mitsugumin72
Gene Aliases: CMT2K; CMYO25; ENSMUSG00000054314; JP-1; JP1; JPH1
UniProt ID: (Human) Q9HDC5, (Mouse) Q9ET80
Entrez Gene ID: (Human) 56704, (Mouse) 57339, (Rat) 297748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support