Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VQAASPERA GWTEALRAAV AELRAGAVVA VPTDTLYGLA CAASCSAALR AVYRLKGRSE AKPLAVCLGR VADVYRYCRV RVPEGLLKDL LPGPVTLVME RSEELNKDLN PFTPLVGIRI PDHAFMQDLA QMFEGPLALT SANLSSQASS LNVEEFQDLW PQLSLVIDGG QIGDGQSPEC RLGSTVVDLS VPGKFGIIRP GCALESTTAI LQQKYGLLPS HASYL (56-279 aa encoded by B C080186)
YrdC (yrdC domain containing protein), also known as IRIP (ischemia/reperfusion-inducible protein homolog), SUA5 or DRIP3 (dopamine receptor-interacting protein 3), is a 279 amino acid ubiquitously expressed protein found at highest levels in brain, liver and pancreas. A member of the SUA5 family, yrdC is involved in certain aspects of transporter activity, such as the regulation of efflux transporter activity and cargo assembly. YrdC is a peripheral membrane protein that contains one yrdC-like domain, interacts with RSC1A1 and localizes to mitochondrial and plasma membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: contains SUA5/yrdC/yciO-like domain; dopamine receptor interacting protein 3; Dopamine receptor-interacting protein 3; hIRIP; ischemia/reperfusion inducible protein; Ischemia/reperfusion-inducible protein homolog; Threonylcarbamoyl-AMP synthase; unnamed protein product; yrdC domain containing; yrdC domain-containing protein, mitochondrial; yrdC N(6)-threonylcarbamoyltransferase domain containing
Gene Aliases: DRIP3; GAMOS10; IRIP; SUA5; YRDC
UniProt ID: (Human) Q86U90
Entrez Gene ID: (Human) 79693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support