Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • N-cadherin Antibodies

          Invitrogen

          N-cadherin Monoclonal Antibody (3B9)

          Advanced Verification
          85 Published Figures
          303 References
          View all (112) N-cadherin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite N-cadherin Monoclonal Antibody (3B9)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (85)
          • Advanced Verification (2)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 93

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          N-cadherin Antibody (33-3900) in ICC/IF

          Immunofluorescent analysis of N-Cadherin was done on 70% confluent log phase Caco-2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes; permeabilized with 0.25% Triton™ X-100 for 10 minutes followed by blocking with 5% BSA for 1 hour at room temperature. The cells were incubated with N-Cadherin Mouse Monoclonal Antibody (Product # 33-3900) at 1 µg - 2 µg in 1% BSA and incubated for 3 hours ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          N-cadherin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          N-cadherin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunocytochemistry (ICC/IF)
          N-cadherin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Immunohistochemistry (IHC)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot, Flow Cytometry (WB, Flow)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody in Western Blot (WB)
          N-cadherin Antibody
          N-cadherin Antibody

          Product Details

          33-3900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 112 publications 112 publications

          Immunohistochemistry (IHC)

          -
          View 81 publications 81 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-5 µg/mL
          View 28 publications 28 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 9 publications 9 publications

          Immunocytochemistry (ICC/IF)

          1-3 µg/mL
          View 47 publications 47 publications

          Flow Cytometry (Flow)

          -
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          3-5 µg
          View 5 publications 5 publications

          Miscellaneous PubMed (Misc)

          -
          View 18 publications 18 publications
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Pig, Rat

          Published species

          Avian, Dog, Hamster, Horse, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Rhesus monkey, Tag, Zebrafish

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3B9

          Immunogen

          Recombinant domain corresponding to the intracellular domain of chicken N-Cadherin

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2313779

          Product Specific Information

          This antibody is specific for N-cadherin and does not cross-react with other cadherin family members including P- and E-cadherins.



          For IHC with this antibody use formalin-fixed, paraffin-embedded tissue and heat-induced epitope retrieval.

          Target Information

          N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; Cadherin-N; Cadherin2; calcium-dependent adhesion protein, neuronal; CD325; CDw325; CDw325 antigen; N-cadherin; N-cadherin 1; Neural cadherin; neural calcium-dependent cell adhesion molecule; unnamed protein product

          View more View less

          Gene Aliases: ACOGS; ADHD8; ARVD14; cadherin-2; CD325; CDH2; CDHN; CDw325; N-cadherin; NCAD

          View more View less

          UniProt ID: (Human) P19022, (Chicken) P10288, (Mouse) P15116, (Rat) Q9Z1Y3

          View more View less

          Entrez Gene ID: (Pig) 100515322, (Human) 1000, (Chicken) 414745, (Mouse) 12558, (Rat) 83501

          View more View less

          Function(s)
          RNA binding calcium ion binding protein binding beta-catenin binding enzyme binding protein kinase binding protein phosphatase binding identical protein binding alpha-catenin binding gamma-catenin binding cadherin binding metal ion binding extracellular matrix binding protein complex binding nitric-oxide synthase binding cadherin
          Process(es)
          cell morphogenesis type B pancreatic cell development cell-cell junction assembly cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules synapse assembly glial cell differentiation neural crest cell development calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules cell migration telencephalon development cerebral cortex development adherens junction organization detection of muscle stretch positive regulation of MAPK cascade cell-cell adhesion mediated by cadherin blood vessel morphogenesis brain morphogenesis homeostasis of number of cells striated muscle cell differentiation radial glial cell differentiation neuroepithelial cell differentiation regulation of oligodendrocyte progenitor proliferation protein localization to plasma membrane negative regulation of canonical Wnt signaling pathway mesenchymal cell migration synaptic vesicle clustering neuroligin clustering involved in postsynaptic membrane assembly neuronal stem cell population maintenance cell-cell adhesion regulation of postsynaptic density protein 95 clustering positive regulation of synaptic vesicle clustering cartilage condensation neural tube formation brain development negative regulation of cell migration neural crest cell delamination regulation of axonogenesis regulation of synaptic transmission, glutamatergic limb development trigeminal ganglion formation trigeminal ganglion structural organization cell-cell adhesion via plasma-membrane adhesion molecules cell aggregation regulation of signal transduction single organismal cell-cell adhesion regulation of myelination regulation of protein localization regulation of Rho protein signal transduction protein heterooligomerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline